Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc5a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053521.1 | NP_445973.1 | 9 | 1815 | ||
XM_006244251.2 | XP_006244313.1 | 9 | 1918 | |||
XM_006244253.2 | XP_006244315.1 | 7 | 1462 | |||
XM_008766774.1 | XP_008764996.1 | 9 | 1966 | |||
XM_008766775.1 | XP_008764997.1 | 9 | 1915 | |||
GenBank mRNA | AB030947.1 | - | 9 | 1815 |