Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006908.4 | NP_008839.2 | 5 | 673 | ||
NM_018890.3 | NP_061485.1 | 6 | 730 | |||
GenBank mRNA | AB451238.1 | - | 5 | 432 | ||
AB451363.1 | - | 5 | 432 | |||
AF136373.1 | - | 6 | 489 | |||
AF498964.1 | - | 5 | 432 | |||
AJ132694.1 | - | 6 | 489 | |||
AK054993.1 | - | 5 | 648 | |||
AK122742.1 | - | 1 | 1576 | |||
AK291806.1 | - | 5 | 629 | |||
AK291815.1 | - | 6 | 676 | |||
AY279384.1 | - | 5 | 590 | |||
BC004247.1 | - | 5 | 570 | 3610745 | ||
BC050687.1 | - | 5 | 620 | 6149377 | ||
BC107748.1 | - | 5 | 635 | 6699945 | ||
BQ689587.1 | - | 6 | 689 | |||
BT007121.1 | - | 5 | 432 | |||
M29870.1 | - | 5 | 432 | |||
M31467.1 | - | 5 | 432 |