Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK10 SIRNA Assays ›
Gene Symbol:
MAPK10
Gene Name:
mitogen-activated protein kinase 10
Gene Aliases:
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
Chromosome Location:
Chr.4: 86012296 - 86594117 on Build GRCh38
UniGene ID:
Hs.125503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
VHS40699
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318067.1
NP_001304996.1
7
775
NM_001318068.1
NP_001304997.1
7
870
NM_001318069.1
NP_001304998.1
8
1340
NM_002753.4
NP_002744.1
8
876
NM_138980.3
NP_620446.1
8
1367
NM_138982.3
NP_620448.1
8
1340
XM_005263129.2
XP_005263186.1
8
1437
XM_005263130.2
XP_005263187.1
8
833
XM_005263131.2
XP_005263188.1
8
1234
XM_005263135.3
XP_005263192.1
8
1234
XM_006714268.2
XP_006714331.1
8
1107
XM_006714269.2
XP_006714332.1
8
1234
XM_011532117.2
XP_011530419.1
8
940
XM_011532118.2
XP_011530420.1
8
1033
XM_011532120.2
XP_011530422.1
8
1003
XM_011532121.2
XP_011530423.1
8
869
XM_017008419.1
XP_016863908.1
9
1130
XM_017008420.1
XP_016863909.1
8
1564
XM_017008421.1
XP_016863910.1
8
1160
XM_017008422.1
XP_016863911.1
6
660
XM_017008423.1
XP_016863912.1
8
852
XM_017008424.1
XP_016863913.1
8
1187
XM_017008425.1
XP_016863914.1
8
860
XM_017008426.1
XP_016863915.1
8
1188
XM_017008427.1
XP_016863916.1
8
1261
XM_017008428.1
XP_016863917.1
8
1003
XM_017008429.1
XP_016863918.1
8
1564
XM_017008430.1
XP_016863919.1
8
942
XM_017008431.1
XP_016863920.1
8
1160
XM_017008432.1
XP_016863921.1
8
833
XM_017008433.1
XP_016863922.1
8
1261
XM_017008434.1
XP_016863923.1
8
1217
XM_017008435.1
XP_016863924.1
8
1107
XM_017008436.1
XP_016863925.1
8
1591
XM_017008437.1
XP_016863926.1
8
869
XM_017008438.1
XP_016863927.1
8
860
XM_017008439.1
XP_016863928.1
8
1188
XM_017008440.1
XP_016863929.1
8
1261
XM_017008441.1
XP_016863930.1
8
1261
XM_017008442.1
XP_016863931.1
10
1582
XM_017008443.1
XP_016863932.1
10
1233
XM_017008444.1
XP_016863933.1
10
1555
XM_017008445.1
XP_016863934.1
7
1064
XM_017008446.1
XP_016863935.1
5
1756
XM_017008447.1
XP_016863936.1
7
1202
XM_017008448.1
XP_016863937.1
7
1175
XM_017008449.1
XP_016863938.1
7
1202
XM_017008450.1
XP_016863939.1
7
1175
XM_017008451.1
XP_016863940.1
7
1001
XM_017008452.1
XP_016863941.1
7
1202
XM_017008453.1
XP_016863942.1
6
660
XM_017008454.1
XP_016863943.1
8
940
GenBank mRNA
AK057723.1
-
3
662
AK124791.1
-
8
1592
AK225974.1
-
8
921
AK294125.1
-
4
391
BC022492.1
-
8
1173
BC035057.1
-
8
1185
5261221
BC051731.1
-
8
986
4838495
BC065516.1
-
3
337
5721954
U07620.1
-
8
877
U34819.1
-
7
721
U34820.1
-
7
745
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
protein phosphorylation
signal transduction
JNK cascade
JUN phosphorylation
response to light stimulus
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
JUN kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0