Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMAD2 SIRNA Assays ›
          Gene Symbol:
          SMAD2
          Gene Name:
          SMAD family member 2
          Gene Aliases:
          JV18, JV18-1, MADH2, MADR2, hMAD-2, hSMAD2
          Chromosome Location:
          Chr.18: 47833095 - 47931193 on Build GRCh38
          UniGene ID:
          Hs.12253
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID VHS41106
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMAD2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003652.3 NP_001003652.1 7 1149
          NM_001135937.2 NP_001129409.1 6 1059
          NM_005901.5 NP_005892.1 7 1041
          XM_005258259.3 XP_005258316.1 8 1282
          XM_006722451.3 XP_006722514.1 8 1294
          XM_011525984.2 XP_011524286.1 7 638
          XM_011525985.2 XP_011524287.1 9 1341
          XM_017025745.1 XP_016881234.1 9 1380
          XM_017025746.1 XP_016881235.1 7 1192
          XM_017025747.1 XP_016881236.1 8 1267
          XM_017025748.1 XP_016881237.1 9 1353
          XM_017025750.1 XP_016881239.1 8 1242
          GenBank mRNA AF027964.1 - 7 855
          AK299218.1 - 6 755
          AK312963.1 - 6 878
          BC014840.1 - 7 997 4449540
          BC025699.1 - 7 855 5221801
          BT007422.1 - 6 749
          U59911.1 - 7 848
          U65019.1 - 6 785
          U68018.1 - 6 804

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ureteric bud development
          in utero embryonic development
          endoderm formation
          mesoderm formation
          transcription from RNA polymerase II promoter
          transforming growth factor beta receptor signaling pathway
          common-partner SMAD protein phosphorylation
          SMAD protein complex assembly
          zygotic specification of dorsal/ventral axis
          gastrulation
          negative regulation of cell proliferation
          response to glucose
          post-embryonic development
          anterior/posterior pattern specification
          positive regulation of epithelial to mesenchymal transition
          regulation of transforming growth factor beta receptor signaling pathway
          signal transduction involved in regulation of gene expression
          insulin secretion
          lung development
          negative regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of BMP signaling pathway
          pancreas development
          primary miRNA processing
          activin receptor signaling pathway
          somatic stem cell population maintenance
          organ growth
          intracellular signal transduction
          nodal signaling pathway
          cell fate commitment
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          paraxial mesoderm morphogenesis
          embryonic foregut morphogenesis
          embryonic cranial skeleton morphogenesis
          regulation of binding
          palate development
          pericardium development
          SMAD protein signal transduction
          response to cholesterol
          positive regulation of nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transforming growth factor beta receptor binding
          protein binding
          transcription factor binding
          phosphatase binding
          transforming growth factor beta receptor, pathway-specific cytoplasmic mediator activity
          ubiquitin protein ligase binding
          activating transcription factor binding
          type I transforming growth factor beta receptor binding
          enhancer binding
          protein homodimerization activity
          SMAD binding
          metal ion binding
          protein heterodimerization activity
          co-SMAD binding
          I-SMAD binding
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0