Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRKCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316329.1 | NP_001303258.1 | 12 | 1638 | ||
NM_002739.4 | NP_002730.1 | 12 | 1638 | |||
GenBank mRNA | AF345987.1 | - | 12 | 1522 | ||
AK296412.1 | - | 12 | 1646 | |||
AK296434.1 | - | 12 | 1625 | |||
AK303741.1 | - | 10 | 1045 | |||
AK312302.1 | - | 12 | 1600 | |||
BC047876.1 | - | 12 | 1513 | 5764695 | ||
Z15114.1 | - | 8 | 846 |