Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other H19 SIRNA Assays ›
Gene Symbol:
H19
Gene Name:
H19, imprinted maternally expressed transcript (non-protein coding)
Gene Aliases:
ASM, ASM1, D11S813E
Chromosome Location:
Chr.1: 215744404 - 215747080 on Build Rnor_6.0
UniGene ID:
Rn.6171
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
n244250
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: H19
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027324.1
3
1538
GenBank mRNA
BC099104.1
-
3
1459
BC160921.1
-
3
1272
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
DY321380.1
-
405
Chr. - - [-] on Build
GenBankmRNA
EV779271.1
-
359
Chr. - - [-] on Build
GenBankmRNA
DY316134.1
-
403
Chr. - - [-] on Build
GenBankmRNA
CB784671.1
-
408
Chr. - - [-] on Build
GenBankmRNA
EV768573.1
-
454
Chr. - - [-] on Build
GenBankmRNA
CB717003.1
-
117
Chr. - - [-] on Build
GenBankmRNA
DY319268.1
-
561
Chr. - - [-] on Build
GenBankmRNA
EV762735.1
-
624
Chr. - - [-] on Build
GenBankmRNA
CB810789.1
-
391
Chr. - - [-] on Build
GenBankmRNA
EB476291.1
-
426
Chr. - - [-] on Build
GenBankmRNA
CB732685.1
-
457
Chr. - - [-] on Build
GenBankmRNA
CV117164.1
-
585
Chr. - - [-] on Build
GenBankmRNA
EV764531.1
-
733
Chr. - - [-] on Build
GenBankmRNA
DY309618.1
-
410
Chr. - - [-] on Build
GenBankmRNA
CB787770.1
-
143
Chr. - - [-] on Build
GenBankmRNA
DY311398.1
-
403
Chr. - - [-] on Build
GenBankmRNA
DY319936.1
-
423
Chr. - - [-] on Build
GenBankmRNA
EV777202.1
-
617
Chr. - - [-] on Build
GenBankmRNA
FN804080.1
-
104
Chr. - - [-] on Build
GenBankmRNA
CB778212.1
-
90
Chr. - - [-] on Build
GenBankmRNA
FN804109.1
-
464
Chr. - - [-] on Build
GenBankmRNA
DY308976.1
-
410
Chr. - - [-] on Build
GenBankmRNA
CB745847.1
-
253
Chr. - - [-] on Build
GenBankmRNA
CB788342.1
-
185
Chr. - - [-] on Build
GenBankmRNA
FN804104.1
-
653
Chr. - - [-] on Build
GenBankmRNA
CV103029.1
-
733
Chr. - - [-] on Build
GenBankmRNA
EV763246.1
-
436
Chr. - - [-] on Build
GenBankmRNA
CB776874.1
-
239
Chr. - - [-] on Build
GenBankmRNA
DY316528.1
-
393
Chr. - - [-] on Build
GenBankmRNA
EV780159.1
-
777
Chr. - - [-] on Build
GenBankmRNA
DY309786.1
-
410
Chr. - - [-] on Build
GenBankmRNA
EV770822.1
-
760
Chr. - - [-] on Build
GenBankmRNA
EV765620.1
-
703
Chr. - - [-] on Build
GenBankmRNA
CB802494.1
-
326
Chr. - - [-] on Build
GenBankmRNA
CB798837.1
-
381
Chr. - - [-] on Build
GenBankmRNA
CB730348.1
-
460
Chr. - - [-] on Build
GenBankmRNA
EV779439.1
-
399
Chr. - - [-] on Build
GenBankmRNA
DY315928.1
-
340
Chr. - - [-] on Build
GenBankmRNA
CB737744.1
-
275
Chr. - - [-] on Build
GenBankmRNA
DY318779.1
-
410
Chr. - - [-] on Build
GenBankmRNA
FN804083.1
-
125
Chr. - - [-] on Build
GenBankmRNA
DY318792.1
-
576
Chr. - - [-] on Build
GenBankmRNA
EV775200.1
-
611
Chr. - - [-] on Build
GenBankmRNA
DY309627.1
-
221
Chr. - - [-] on Build
GenBankmRNA
EV774568.1
-
782
Chr. - - [-] on Build
GenBankmRNA
DY313825.1
-
552
Chr. - - [-] on Build
GenBankmRNA
CB729442.1
-
347
Chr. - - [-] on Build
GenBankmRNA
FM029648.1
-
410
Chr. - - [-] on Build
GenBankmRNA
CV103161.1
-
217
Chr. - - [-] on Build
GenBankmRNA
CV120053.1
-
393
Chr. - - [-] on Build
GenBankmRNA
CV115681.1
-
551
Chr. - - [-] on Build
GenBankmRNA
EV768581.1
-
629
Chr. - - [-] on Build
GenBankmRNA
EV780708.1
-
784
Chr. - - [-] on Build
GenBankmRNA
FN804112.1
-
236
Chr. - - [-] on Build
GenBankmRNA
CB744019.1
-
360
Chr. - - [-] on Build
GenBankmRNA
EV769573.1
-
651
Chr. - - [-] on Build
GenBankmRNA
CV113239.1
-
626
Chr. - - [-] on Build
GenBankmRNA
CB741501.1
-
268
Chr. - - [-] on Build
GenBankmRNA
FN804108.1
-
384
Chr. - - [-] on Build
GenBankmRNA
EV767862.1
-
111
Chr. - - [-] on Build
GenBankmRNA
EV770990.1
-
697
Chr. - - [-] on Build
GenBankmRNA
DY309583.1
-
406
Chr. - - [-] on Build
GenBankmRNA
DY316240.1
-
552
Chr. - - [-] on Build
GenBankmRNA
CB758044.1
-
224
Chr. - - [-] on Build
GenBankmRNA
EV764754.1
-
613
Chr. - - [-] on Build
GenBankmRNA
EV768815.1
-
640
Chr. - - [-] on Build
GenBankmRNA
CV112487.1
-
617
Chr. - - [-] on Build
GenBankmRNA
EV777316.1
-
344
Chr. - - [-] on Build
GenBankmRNA
FN804111.1
-
417
Chr. - - [-] on Build
GenBankmRNA
DY309155.1
-
550
Chr. - - [-] on Build
GenBankmRNA
DY317598.1
-
423
Chr. - - [-] on Build
GenBankmRNA
CB616745.1
-
180
Chr. - - [-] on Build
GenBankmRNA
DY319344.1
-
555
Chr. - - [-] on Build
GenBankmRNA
CB707294.1
-
359
Chr. - - [-] on Build
GenBankmRNA
CV105643.1
-
419
Chr. - - [-] on Build
GenBankmRNA
CB789056.1
-
96
Chr. - - [-] on Build
GenBankmRNA
EV765723.1
-
398
Chr. - - [-] on Build
GenBankmRNA
CA510644.1
-
586
Chr. - - [-] on Build
GenBankmRNA
DY308870.1
-
364
Chr. - - [-] on Build
GenBankmRNA
FQ117437.1
-
688
Chr. - - [-] on Build
GenBankmRNA
CV111815.1
-
551
Chr. - - [-] on Build
GenBankmRNA
CV108855.1
-
409
Chr. - - [-] on Build
GenBankmRNA
EV770283.1
-
461
Chr. - - [-] on Build
GenBankmRNA
DY319332.1
-
631
Chr. - - [-] on Build
GenBankmRNA
CB741497.1
-
102
Chr. - - [-] on Build
GenBankmRNA
FQ116831.1
-
625
Chr. - - [-] on Build
GenBankmRNA
EV764569.1
-
726
Chr. - - [-] on Build
GenBankmRNA
DY319044.1
-
404
Chr. - - [-] on Build
GenBankmRNA
CV104032.1
-
429
Chr. - - [-] on Build
GenBankmRNA
EV767032.1
-
654
Chr. - - [-] on Build
GenBankmRNA
DY310815.1
-
566
Chr. - - [-] on Build
GenBankmRNA
FN804110.1
-
486
Chr. - - [-] on Build
GenBankmRNA
CB786289.1
-
362
Chr. - - [-] on Build
GenBankmRNA
CB732670.1
-
284
Chr. - - [-] on Build
GenBankmRNA
CB751384.1
-
207
Chr. - - [-] on Build
GenBankmRNA
CV105546.1
-
491
Chr. - - [-] on Build
GenBankmRNA
CB758045.1
-
354
Chr. - - [-] on Build
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
liver development
skeletal muscle tissue development
response to wounding
response to nutrient levels
response to lipid
response to peptide hormone
response to steroid hormone
response to glucocorticoid
mesenchymal to epithelial transition
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0