Search Thermo Fisher Scientific
Gene Symbol: Smok4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030763.1 | 2 | 1777 | |||
GenBank mRNA | AK029728.1 | - | 2 | 1778 | ||
BC125398.1 | - | 1 | 1336 | |||
BC132092.1 | - | 1 | 1336 |