Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG3 SIRNA Assays ›
          Gene Symbol:
          Snhg3
          Gene Name:
          small nucleolar RNA host gene 3
          Gene Aliases:
          Rnu17d, U17HG
          Chromosome Location:
          Chr.4: 132351933 - 132353686 on Build GRCm38
          UniGene ID:
          Mm.486919
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n253535

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Snhg3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003270.2
          3 180
          GenBank mRNA AJ006837.1 - LIT1313
          U17HG
          3 127
          AK076188.1 -
          3 128
          BC100513.1 -
          3 173
          CN834544.1 -
          3 183
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY118837.1 - 3 164 Chr. 4 - 132352035 - 132353668 [-] on Build GRCm38
          GenBankmRNA EL959114.1 - 3 172 Chr. 4 - 132351934 - 132353651 [-] on Build GRCm38
          GenBankmRNA CN841655.1 - 3 176 Chr. 4 - 132351934 - 132353679 [-] on Build GRCm38
          GenBankmRNA DV069876.1 - 196 Chr. - - [-] on Build
          GenBankmRNA DV069651.1 - 147 Chr. - - [-] on Build
          GenBankmRNA DV061498.1 - 225 Chr. - - [-] on Build
          GenBankmRNA CA460116.1 - 217 Chr. - - [-] on Build
          GenBankmRNA DV062701.1 - 248 Chr. - - [-] on Build
          GenBankmRNA AI787530.1 - 3 126 Chr. 4 - 132351934 - 132353632 [-] on Build GRCm38
          GenBankmRNA CJ216122.1 - 2 142 Chr. 4 - 132351934 - 132352750 [-] on Build GRCm38
          GenBankmRNA CA456802.1 - 194 Chr. - - [-] on Build
          GenBankmRNA BY144490.1 - 3 124 Chr. 4 - 132351966 - 132353630 [-] on Build GRCm38
          GenBankmRNA BY121084.1 - 3 163 Chr. 4 - 132352035 - 132353668 [-] on Build GRCm38
          GenBankmRNA BB605517.1 - 3 130 Chr. 4 - 132352034 - 132353634 [-] on Build GRCm38
          GenBankmRNA DV072144.1 - 147 Chr. - - [-] on Build
          GenBankmRNA BE198161.1 - 3 154 Chr. 4 - 132352117 - 132353660 [-] on Build GRCm38
          GenBankmRNA CA492396.1 - 164 Chr. - - [-] on Build
          GenBankmRNA AJ006836.1 - 1 1427 Chr. - - [-] on Build
          GenBankmRNA BY713659.1 - 3 122 Chr. 4 - 132351933 - 132353628 [-] on Build GRCm38
          GenBankmRNA AI788223.1 - 3 126 Chr. 4 - 132351934 - 132353632 [-] on Build GRCm38
          GenBankmRNA BY792896.1 - 3 158 Chr. 4 - 132348014 - 132353663 [-] on Build GRCm38
          GenBankmRNA BY579581.1 - 2 168 Chr. 4 - 132351934 - 132352776 [-] on Build GRCm38
          GenBankmRNA BY121260.1 - 3 163 Chr. 4 - 132352022 - 132353668 [-] on Build GRCm38
          GenBankmRNA DV041333.1 - 222 Chr. - - [-] on Build
          GenBankmRNA CA786577.1 - 176 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0