Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BTBD19 SIRNA Assays ›
          Gene Symbol:
          Btbd19
          Gene Name:
          BTB (POZ) domain containing 19
          Gene Aliases:
          9530048O09Rik, BB115071
          Chromosome Location:
          Chr.4: 117119218 - 117125725 on Build GRCm38
          UniGene ID:
          Mm.446227
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n254132

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Btbd19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024078.1 7 1060
          GenBank mRNA AK020599.1 - 2 205
          AK141215.1 - 2 292
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY722133.1 - 2 205 Chr. 4 - 117120340 - 117121275 [-] on Build GRCm38
          GenBankmRNA CJ142189.1 - 2 421 Chr. 4 - 117120706 - 117121265 [-] on Build GRCm38
          GenBankmRNA BY022447.1 - 3 310 Chr. 4 - 117120651 - 117122334 [-] on Build GRCm38
          GenBankmRNA BY092682.1 - 2 232 Chr. 4 - 117120584 - 117121303 [-] on Build GRCm38
          GenBankmRNA BB651729.1 - 2 292 Chr. 4 - 117120461 - 117122385 [-] on Build GRCm38
          GenBankmRNA BY724455.1 - 2 232 Chr. 4 - 117120301 - 117121304 [-] on Build GRCm38
          GenBankmRNA CJ071803.1 - 4 439 Chr. 4 - 117120710 - 117122330 [-] on Build GRCm38
          GenBankmRNA NM_001033187.1 - 292 Chr. - - [-] on Build
          GenBankmRNA AA000417.1 - 2 226 Chr. 4 - 117120514 - 117121297 [-] on Build GRCm38
          GenBankmRNA BY026486.1 - 3 312 Chr. 4 - 117120679 - 117122334 [-] on Build GRCm38
          GenBankmRNA AA034670.1 - 2 224 Chr. 4 - 117120458 - 117121297 [-] on Build GRCm38
          GenBankmRNA BY019083.1 - 3 270 Chr. 4 - 117120664 - 117122294 [-] on Build GRCm38
          GenBankmRNA CJ154914.1 - 2 230 Chr. 4 - 117120517 - 117121302 [-] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0