Search
Search

9330175E14Rik| Gene Symbol: 9330175E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015514.2 | 1 | 17 | |||
| GenBank mRNA | AK053303.1 | - | 1 | 19 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB664790.1 | - | 1 | 19 | Chr. 8 - 94434039 - 94435105 [-] on Build GRCm38 | |