Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WAC SIRNA Assays ›
          Gene Symbol:
          Wac
          Gene Name:
          WW domain containing adaptor with coiled-coil
          Gene Aliases:
          1110067P07Rik, A230035H12Rik, AI256735, AI256776, Wwp4
          Chromosome Location:
          Chr.18: 7868459 - 7929028 on Build GRCm38
          UniGene ID:
          Mm.204524
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n255339

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Wac
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK156780.1 - 1 167
          AK172416.1 - 1 167
          BC020146.1 - 1 155
          CB249484.1 - 1 158
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY151258.1 - 1 165 Chr. 18 - 7868852 - 7871609 [+] on Build GRCm38
          GenBankmRNA AI930409.1 - 1 184 Chr. 18 - 7868832 - 7871713 [+] on Build GRCm38
          GenBankmRNA BY140407.1 - 1 165 Chr. 18 - 7868852 - 7871638 [+] on Build GRCm38
          GenBankmRNA BY784849.1 - 1 161 Chr. 18 - 7868856 - 7871620 [+] on Build GRCm38
          GenBankmRNA BY265457.1 - 1 169 Chr. 18 - 7868847 - 7871717 [+] on Build GRCm38
          GenBankmRNA BY292420.1 - 1 164 Chr. 18 - 7868852 - 7871639 [+] on Build GRCm38
          GenBankmRNA CA979433.1 - 152 Chr. - - [-] on Build
          GenBankmRNA CN688953.1 - 1 157 Chr. 18 - 7868859 - 7905490 [+] on Build GRCm38
          GenBankmRNA CJ141540.1 - 1 173 Chr. 18 - 7868845 - 7871684 [+] on Build GRCm38
          GenBankmRNA BY296369.1 - 1 164 Chr. 18 - 7868852 - 7871667 [+] on Build GRCm38
          GenBankmRNA BF139979.1 - 1 158 Chr. 18 - 7868859 - 7905450 [+] on Build GRCm38
          GenBankmRNA BY752036.1 - 1 164 Chr. 18 - 7868852 - 7905513 [+] on Build GRCm38
          GenBankmRNA BY751648.1 - 1 164 Chr. 18 - 7868852 - 7905513 [+] on Build GRCm38
          GenBankmRNA AI747996.1 - 1 163 Chr. 18 - 7868851 - 7905445 [+] on Build GRCm38
          GenBankmRNA CF897039.1 - 1 150 Chr. 18 - 7868866 - 7905475 [+] on Build GRCm38
          GenBankmRNA BU053437.1 - 1 155 Chr. 18 - 7868861 - 7906011 [+] on Build GRCm38
          GenBankmRNA CF723514.1 - 1 121 Chr. 18 - 7868894 - 7905515 [+] on Build GRCm38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0