Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PISD-PS3 SIRNA Assays ›
          Gene Symbol:
          Pisd-ps3
          Gene Name:
          phosphatidylserine decarboxylase, pseudogene 3
          Gene Aliases:
          4933439C20Rik
          Chromosome Location:
          Chr.Un|NT_187064: 52674 - 59689 on Build GRCm38
          UniGene ID:
          Mm.247625
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n255839

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pisd-ps3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003518.2 2 360
          GenBank mRNA AK005333.1 - 464
          AK017116.1 - 2 369
          AK034517.1 - 2 1017
          AK036319.1 - 2 290
          AK081907.1 - 2 337
          AK144226.1 - 1 19
          AK161130.1 - 2 360
          AK162734.1 - 2 1493
          AK168892.1 - 2 337
          AK169597.1 - 1 1103
          AK169755.1 - 1 1103
          AK183211.1 - 1 13
          AK185609.1 - 1 272
          AK185737.1 - 1 278
          AK188188.1 - 1 13
          BC048401.1 - 1 390
          BC059938.1 - 1 266
          BC083061.1 - 1 108
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CA319555.1 - 2 377 Chr. Un|NT_187064.1 - 55497 - 59619 [-] on Build GRCm38
          GenBankmRNA AU080934.1 - 375 Chr. - - [-] on Build
          GenBankmRNA BQ945049.1 - 1 265 Chr. Un|NT_187064.1 - 55117 - 58837 [-] on Build GRCm38
          GenBankmRNA CN672583.1 - 1 113 Chr. Un|NT_187064.1 - 55456 - 58688 [-] on Build GRCm38
          GenBankmRNA CJ171901.1 - 2 333 Chr. Un|NT_187064.1 - 57106 - 59663 [-] on Build GRCm38
          GenBankmRNA BQ044418.1 - 1 34 Chr. Un|NT_187064.1 - 55302 - 58609 [-] on Build GRCm38
          GenBankmRNA BY739907.1 - 2 337 Chr. Un|NT_187064.1 - 55575 - 59667 [-] on Build GRCm38
          GenBankmRNA BB619177.1 - 2 337 Chr. Un|NT_187064.1 - 55615 - 59667 [-] on Build GRCm38
          GenBankmRNA BY255235.1 - 2 335 Chr. Un|NT_187064.1 - 57004 - 59667 [-] on Build GRCm38
          GenBankmRNA BQ745548.1 - 2 305 Chr. Un|NT_187064.1 - 55444 - 59637 [-] on Build GRCm38
          GenBankmRNA CA531192.1 - 1 80 Chr. Un|NT_187064.1 - 55449 - 58655 [-] on Build GRCm38
          GenBankmRNA CJ175330.1 - 2 338 Chr. Un|NT_187064.1 - 57039 - 59667 [-] on Build GRCm38
          GenBankmRNA CA318015.1 - 1 113 Chr. Un|NT_187064.1 - 54728 - 58687 [-] on Build GRCm38
          GenBankmRNA BB649681.1 - 2 337 Chr. Un|NT_187064.1 - 55570 - 59667 [-] on Build GRCm38
          GenBankmRNA BQ179332.1 - 1 33 Chr. Un|NT_187064.1 - 55157 - 58608 [-] on Build GRCm38
          GenBankmRNA BF140154.1 - 100 Chr. - - [-] on Build
          GenBankmRNA BQ443975.1 - 1 111 Chr. Un|NT_187064.1 - 54776 - 58677 [-] on Build GRCm38
          GenBankmRNA BP766142.1 - 1 448 Chr. Un|NT_187064.1 - 25388 - 58997 [-] on Build GRCm38
          GenBankmRNA CA533423.1 - 1 188 Chr. Un|NT_187064.1 - 57025 - 58768 [-] on Build GRCm38
          GenBankmRNA BY273730.1 - 2 338 Chr. Un|NT_187064.1 - 57051 - 59667 [-] on Build GRCm38
          GenBankmRNA AU066585.1 - 2 354 Chr. 11 - 3124028 - 3127613 [+] on Build GRCm38
          GenBankmRNA CX207012.1 - 2 359 Chr. Un|NT_187064.1 - 55530 - 59690 [-] on Build GRCm38
          GenBankmRNA CK620355.1 - 1 127 Chr. Un|NT_187064.1 - 53068 - 58702 [-] on Build GRCm38
          GenBankmRNA BI697592.1 - 1 111 Chr. Un|NT_187064.1 - 55243 - 58686 [-] on Build GRCm38
          GenBankmRNA CN671160.1 - 1 171 Chr. Un|NT_187064.1 - 55467 - 58748 [-] on Build GRCm38
          GenBankmRNA BM951557.1 - 1 107 Chr. Un|NT_187064.1 - 55277 - 58681 [-] on Build GRCm38
          GenBankmRNA CN681062.1 - 1 111 Chr. Un|NT_187064.1 - 25151 - 58686 [-] on Build GRCm38
          GenBankmRNA AA607577.1 - 1 84 Chr. Un|NT_187064.1 - 57059 - 58659 [-] on Build GRCm38
          GenBankmRNA BY722385.1 - 2 332 Chr. Un|NT_187064.1 - 25366 - 59664 [-] on Build GRCm38
          GenBankmRNA CX237225.1 - 2 407 Chr. Un|NT_187064.1 - 55545 - 59731 [-] on Build GRCm38
          GenBankmRNA BQ443910.1 - 1 252 Chr. Un|NT_187064.1 - 55206 - 58826 [-] on Build GRCm38
          GenBankmRNA BY716795.1 - 2 363 Chr. Un|NT_187064.1 - 56853 - 59695 [-] on Build GRCm38
          GenBankmRNA BU611278.1 - 2 474 Chr. Un|NT_187064.1 - 55658 - 59729 [-] on Build GRCm38
          GenBankmRNA CB519629.1 - 289 Chr. - - [-] on Build
          GenBankmRNA CN680240.1 - 1 111 Chr. Un|NT_187064.1 - 55481 - 58686 [-] on Build GRCm38
          GenBankmRNA CK618982.1 - 2 282 Chr. Un|NT_187064.1 - 57030 - 59613 [-] on Build GRCm38
          GenBankmRNA CF537165.1 - 116 Chr. - - [-] on Build
          GenBankmRNA CA315346.1 - 2 478 Chr. Un|NT_187064.1 - 55552 - 59792 [-] on Build GRCm38
          GenBankmRNA CJ174094.1 - 2 338 Chr. Un|NT_187064.1 - 57068 - 59667 [-] on Build GRCm38
          GenBankmRNA BF141965.1 - 99 Chr. - - [-] on Build
          GenBankmRNA CB245085.1 - 132 Chr. - - [-] on Build
          GenBankmRNA AW321782.1 - 1 110 Chr. Un|NT_187064.1 - 55453 - 58685 [-] on Build GRCm38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          decarboxylase

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dt4l6:80/100.66.76.4:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0