Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP2A2 SIRNA Assays ›
          Gene Symbol:
          Atp2a2
          Gene Name:
          ATPase, Ca++ transporting, cardiac muscle, slow twitch 2
          Gene Aliases:
          9530097L16Rik, D5Wsu150e, SERCA2, SERCA2B, Serca2a, mKIAA4195
          Chromosome Location:
          Chr.5: 122453513 - 122502225 on Build GRCm38
          UniGene ID:
          Mm.227583
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n257020

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Atp2a2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027838.1 20, 21 4927
          GenBank mRNA AK081191.1 - 1, 2 564
          AK134659.1 - 1, 2 1096
          AK220538.1 - 20, 21 4876
          BC054531.1 - 20, 21 4586 6406916
          BC054748.1 - 20, 21 4586 6406916

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NR_027839, CO404645, J04022, FQ213049.

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of the force of heart contraction
          transport
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          regulation of muscle contraction
          ER-nucleus signaling pathway
          organelle organization
          regulation of cardiac muscle contraction by calcium ion signaling
          transition between fast and slow fiber
          endoplasmic reticulum calcium ion homeostasis
          positive regulation of endoplasmic reticulum calcium ion concentration
          T-tubule organization
          response to peptide hormone
          negative regulation of heart contraction
          relaxation of cardiac muscle
          sarcoplasmic reticulum calcium ion transport
          calcium ion transmembrane transport
          regulation of cardiac muscle cell membrane potential
          regulation of cardiac muscle cell action potential involved in regulation of contraction
          regulation of calcium ion-dependent exocytosis of neurotransmitter
          calcium ion transport from cytosol to endoplasmic reticulum
          calcium ion import into sarcoplasmic reticulum
          nucleotide binding
          calcium-transporting ATPase activity
          calcium ion binding
          protein binding
          ATP binding
          protein C-terminus binding
          hydrolase activity
          enzyme binding
          lutropin-choriogonadotropic hormone receptor binding
          S100 protein binding
          metal ion binding
          calcium-transporting ATPase activity involved in regulation of cardiac muscle cell membrane potential

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-lmn4x:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline