Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 1700012D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015573.2 | 2 | 325 | |||
GenBank mRNA | AK005898.1 | - | 2 | 327 | ||
AK047116.1 | - | 1 | 1706 | |||
AK087602.1 | - | 1 | 1695 |