Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: KLKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002948.1 | 5 | 738 | |||
GenBank mRNA | AF355199.1 | - | 2 | 300 | ||
AF464754.1 | - | 2 | 476 | |||
AY533562.1 | - | 5 | 738 | |||
DQ086829.1 | - | 2 | 1145 | |||
DQ211698.1 | - | 5 | 2461 |