Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAHD2CP SIRNA Assays ›
          Gene Symbol:
          FAHD2CP
          Gene Name:
          fumarylacetoacetate hydrolase domain containing 2C, pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 96010551 - 96023136 on Build GRCh38
          UniGene ID:
          Hs.720108
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n258354

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAHD2CP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003698.1 2 207
          GenBank mRNA BI916267.1 - 2 174
          BM790702.1 - 2 207
          CN274361.1 - 2 140
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM845774.1 - 2 267 Chr. 2 - 96010616 - 96022732 [+] on Build GRCh38
          GenBankmRNA BG702382.1 - 2 212 Chr. 2 - 96010551 - 96023249 [+] on Build GRCh38
          GenBankmRNA BM549326.1 - 3 345 Chr. 2 - 96010630 - 96023071 [+] on Build GRCh38
          GenBankmRNA DB498802.1 - 2 165 Chr. 2 - 96010593 - 96022837 [+] on Build GRCh38
          GenBankmRNA HY106469.1 - 4 281 Chr. 2 - 96010611 - 96022714 [+] on Build GRCh38
          GenBankmRNA BG704720.1 - 2 170 Chr. 2 - 96010591 - 96023118 [+] on Build GRCh38
          GenBankmRNA HY004068.1 - 2 163 Chr. 2 - 96010596 - 96022853 [+] on Build GRCh38
          GenBankmRNA BI753039.1 - 2 144 Chr. 2 - 96010614 - 96023380 [+] on Build GRCh38
          GenBankmRNA HY054888.1 - 2 185 Chr. 2 - 96010574 - 96022727 [+] on Build GRCh38
          GenBankmRNA CD673198.1 - 2 164 Chr. 2 - 96010594 - 96022837 [+] on Build GRCh38
          GenBankmRNA BQ067994.1 - 3 413 Chr. 2 - 96010584 - 96023208 [+] on Build GRCh38
          GenBankmRNA DB472255.1 - 2 165 Chr. 2 - 96010593 - 96022819 [+] on Build GRCh38
          GenBankmRNA DB463572.1 - 2 166 Chr. 2 - 96010593 - 96022752 [+] on Build GRCh38
          GenBankmRNA N77563.1 - 2 109 Chr. 2 - 96019619 - 96022878 [+] on Build GRCh38
          GenBankmRNA BM449595.1 - 2 84 Chr. 2 - 96010674 - 96023338 [+] on Build GRCh38
          GenBankmRNA BG714493.1 - 2 150 Chr. 2 - 96010611 - 96023031 [+] on Build GRCh38
          GenBankmRNA DB465674.1 - 2 204 Chr. 2 - 96010554 - 96022698 [+] on Build GRCh38

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: AK005134, AK002291, AK170875, BC030182, AK080868.

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0