Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAHD2CP SIRNA Assays ›
          Gene Symbol:
          FAHD2CP FAHD2A FAHD2B
          Gene Name:
          fumarylacetoacetate hydrolase domain containing 2C, pseudogene
          fumarylacetoacetate hydrolase domain containing 2A
          fumarylacetoacetate hydrolase domain containing 2B
          Gene Aliases:
          -
          CGI-105
          -
          Chromosome Location:
          Chr.2: 96010551 - 96023136 on Build GRCh38
          Chromosome Location:
          Chr.2: 95402691 - 95416702 on Build GRCh38
          Chromosome Location:
          Chr.2: 97080738 - 97094882 on Build GRCh38
          UniGene ID:
          Hs.720108
          Hs.531247
          Hs.567723
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID n258357

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAHD2CP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003698.1 3, 4 387
          GenBank mRNA BI916267.1 - 3, 4 354
          CN274361.1 - 3, 4 320
          Gene Symbol: FAHD2A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011511282.2 XP_011509584.1 7, 8 1808
          XM_011511283.2 XP_011509585.1 8, 9 1185
          XM_011511284.2 XP_011509586.1 8, 9 1314
          XM_011511285.2 XP_011509587.1 8, 9 1182
          XM_011511286.2 XP_011509588.1 7, 8 1068
          XM_011511287.2 XP_011509589.1 7, 8 1179
          XM_011511288.2 XP_011509590.1 7, 8 1808
          Gene Symbol: FAHD2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011510745.2 XP_011509047.1 8, 9 1184
          XM_011510746.2 XP_011509048.1 8, 9 1303
          XM_011510747.2 XP_011509049.1 7, 8 1068
          XM_011510748.2 XP_011509050.1 8, 9 1184
          XM_017003470.1 XP_016858959.1 8, 9 1181
          XM_017003471.1 XP_016858960.1 8, 9 1184
          XM_017003472.1 XP_016858961.1 7, 8 1117
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM845774.1 - 3, 4 447 Chr. 2 - 96010616 - 96022732 [+] on Build GRCh38
          GenBankmRNA BM549326.1 - 4, 5 525 Chr. 2 - 96010630 - 96023071 [+] on Build GRCh38
          GenBankmRNA BG702382.1 - 3, 4 392 Chr. 2 - 96010551 - 96023249 [+] on Build GRCh38
          GenBankmRNA DB498802.1 - 3, 4 345 Chr. 2 - 96010593 - 96022837 [+] on Build GRCh38
          GenBankmRNA BG704720.1 - 3, 4 350 Chr. 2 - 96010591 - 96023118 [+] on Build GRCh38
          GenBankmRNA HY106469.1 - 5, 6 461 Chr. 2 - 96010611 - 96022714 [+] on Build GRCh38
          GenBankmRNA HY004068.1 - 3, 4 343 Chr. 2 - 96010596 - 96022853 [+] on Build GRCh38
          GenBankmRNA BI753039.1 - 3, 4 324 Chr. 2 - 96010614 - 96023380 [+] on Build GRCh38
          GenBankmRNA HY054888.1 - 3, 4 365 Chr. 2 - 96010574 - 96022727 [+] on Build GRCh38
          GenBankmRNA BQ067994.1 - 4, 5 593 Chr. 2 - 96010584 - 96023208 [+] on Build GRCh38
          GenBankmRNA N77563.1 - 3, 4 289 Chr. 2 - 96019619 - 96022878 [+] on Build GRCh38
          GenBankmRNA BM449595.1 - 3, 4 264 Chr. 2 - 96010674 - 96023338 [+] on Build GRCh38
          GenBankmRNA DB465674.1 - 3, 4 384 Chr. 2 - 96010554 - 96022698 [+] on Build GRCh38
          GenBankmRNA CD673198.1 - 3, 4 344 Chr. 2 - 96010594 - 96022837 [+] on Build GRCh38
          GenBankmRNA DB472255.1 - 3, 4 345 Chr. 2 - 96010593 - 96022819 [+] on Build GRCh38
          GenBankmRNA DB463572.1 - 3, 4 345 Chr. 2 - 96010593 - 96022752 [+] on Build GRCh38
          GenBankmRNA BG714493.1 - 3, 4 330 Chr. 2 - 96010611 - 96023031 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          hydrolase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0