Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF542P SIRNA Assays ›
          Gene Symbol:
          ZNF542P
          Gene Name:
          zinc finger protein 542, pseudogene
          Gene Aliases:
          ZNF542
          Chromosome Location:
          Chr.19: 56368099 - 56379828 on Build GRCh38
          UniGene ID:
          Hs.467326
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n258914

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF542P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003127.2 7 609
          NR_024055.2 6 512
          NR_024056.2 6 765
          NR_024057.2 5 693
          NR_033418.1 6 833
          GenBank mRNA AK097128.1 - 5 497
          AK297170.1 - 6 507
          AL833621.1 - 5 462
          BC089419.1 - 5 437
          BX640680.1 - 7 569
          BX647919.1 - 6 568
          DA095148.1 - 6 512
          DA679733.1 - 6 513
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA412445.1 - 5 460 Chr. 19 - 56368354 - 56376957 [+] on Build GRCh38
          GenBankmRNA DC349742.1 - 6 507 Chr. 19 - 56368139 - 56376982 [+] on Build GRCh38
          GenBankmRNA DA760867.1 - 5 478 Chr. 19 - 56368336 - 56377266 [+] on Build GRCh38
          GenBankmRNA DA752878.1 - 5 460 Chr. 19 - 56368354 - 56377029 [+] on Build GRCh38
          GenBankmRNA DA710487.1 - 5 482 Chr. 19 - 56368334 - 56377003 [+] on Build GRCh38
          GenBankmRNA DN991121.1 - 3 233 Chr. 19 - 56373214 - 56377322 [+] on Build GRCh38
          GenBankmRNA DB269839.1 - 5 480 Chr. 19 - 56368336 - 56377006 [+] on Build GRCh38
          GenBankmRNA BP429366.1 - 4 381 Chr. 19 - 56373202 - 56376948 [+] on Build GRCh38
          GenBankmRNA DA950968.1 - 5 495 Chr. 19 - 56368319 - 56377005 [+] on Build GRCh38
          GenBankmRNA BI769075.1 - 6 538 Chr. 19 - 56368348 - 56377184 [+] on Build GRCh38
          GenBankmRNA DA999565.1 - 5 480 Chr. 19 - 56368336 - 56377037 [+] on Build GRCh38
          GenBankmRNA DA031351.1 - 6 551 Chr. 19 - 56368336 - 56376956 [+] on Build GRCh38
          GenBankmRNA DA724793.1 - 5 478 Chr. 19 - 56368336 - 56377008 [+] on Build GRCh38
          GenBankmRNA CB854015.1 - 5 468 Chr. 19 - 56368345 - 56377070 [+] on Build GRCh38
          GenBankmRNA DA947581.1 - 5 479 Chr. 19 - 56368336 - 56377027 [+] on Build GRCh38
          GenBankmRNA DB148486.1 - 5 495 Chr. 19 - 56368319 - 56376989 [+] on Build GRCh38
          GenBankmRNA DA789269.1 - 5 496 Chr. 19 - 56368319 - 56377010 [+] on Build GRCh38
          GenBankmRNA DB004640.1 - 5 480 Chr. 19 - 56368336 - 56377001 [+] on Build GRCh38
          GenBankmRNA DA027434.1 - 5 479 Chr. 19 - 56368336 - 56377039 [+] on Build GRCh38
          GenBankmRNA DB184581.1 - 5 480 Chr. 19 - 56368334 - 56377011 [+] on Build GRCh38
          GenBankmRNA DA660373.1 - 5 478 Chr. 19 - 56368338 - 56377015 [+] on Build GRCh38
          GenBankmRNA DA187598.1 - 5 511 Chr. 19 - 56368303 - 56376954 [+] on Build GRCh38
          GenBankmRNA DA854153.1 - 5 479 Chr. 19 - 56368336 - 56377043 [+] on Build GRCh38
          GenBankmRNA CX760208.1 - 5 466 Chr. 19 - 56368348 - 56377143 [+] on Build GRCh38
          GenBankmRNA DA590960.1 - 5 479 Chr. 19 - 56368336 - 56377024 [+] on Build GRCh38
          GenBankmRNA DA584115.1 - 5 495 Chr. 19 - 56368319 - 56377028 [+] on Build GRCh38
          GenBankmRNA CN310858.1 - 5 474 Chr. 19 - 56368340 - 56377112 [+] on Build GRCh38
          GenBankmRNA DA990420.1 - 5 479 Chr. 19 - 56368336 - 56376990 [+] on Build GRCh38
          GenBankmRNA DA485448.1 - 5 478 Chr. 19 - 56368336 - 56376998 [+] on Build GRCh38
          GenBankmRNA DN998899.1 - 4 318 Chr. 19 - 56373205 - 56377324 [+] on Build GRCh38
          GenBankmRNA AL539099.3 - 5 476 Chr. 19 - 56368348 - 56377469 [+] on Build GRCh38
          GenBankmRNA DA955624.1 - 5 481 Chr. 19 - 56368334 - 56377017 [+] on Build GRCh38
          GenBankmRNA DC371520.1 - 5 435 Chr. 19 - 56368139 - 56376990 [+] on Build GRCh38
          GenBankmRNA DA823197.1 - 5 496 Chr. 19 - 56368319 - 56376992 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0