Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ARMCX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028407.2 | 15 | 3419 | |||
NR_045862.1 | 11 | 2439 | ||||
GenBank mRNA | AK125041.1 | - | 11 | 2237 | ||
CR933662.1 | - | 15 | 3394 |