Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC148413 SIRNA Assays ›
          Gene Symbol:
          LOC148413
          Gene Name:
          uncharacterized LOC148413
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 1399530 - 1402046 on Build GRCh38
          UniGene ID:
          Hs.741357
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n259654

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC148413
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015434.1 3 766
          GenBank mRNA AK024040.1 - 3 740
          AK092572.1 - 1 1259
          AK297832.1 - 3 755
          BC068600.1 - 3 723
          BC080546.1 - 3 722
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI767355.1 - 3 710 Chr. 1 - 1399582 - 1401172 [+] on Build GRCh38
          GenBankmRNA AL528291.3 - 3 896 Chr. 1 - 1399572 - 1401180 [+] on Build GRCh38
          GenBankmRNA BM921447.1 - 3 726 Chr. 1 - 1399572 - 1401278 [+] on Build GRCh38
          GenBankmRNA BQ061816.1 - 3 703 Chr. 1 - 1399594 - 1401252 [+] on Build GRCh38
          GenBankmRNA BX376671.2 - 3 743 Chr. 1 - 1399575 - 1401329 [+] on Build GRCh38
          GenBankmRNA AL548695.3 - 3 686 Chr. 1 - 1399606 - 1401277 [+] on Build GRCh38
          GenBankmRNA BQ070201.1 - 3 719 Chr. 1 - 1399576 - 1401291 [+] on Build GRCh38
          GenBankmRNA BQ058973.1 - 3 702 Chr. 1 - 1399594 - 1401350 [+] on Build GRCh38
          GenBankmRNA BI257793.1 - 636 Chr. - - [-] on Build
          GenBankmRNA DA953265.1 - 360 Chr. - - [-] on Build
          GenBankmRNA BM547793.1 - 3 705 Chr. 1 - 1399591 - 1401261 [+] on Build GRCh38
          GenBankmRNA BQ064163.1 - 702 Chr. - - [-] on Build
          GenBankmRNA AL533370.3 - 3 753 Chr. 1 - 1399561 - 1401295 [+] on Build GRCh38
          GenBankmRNA AL527639.3 - 3 719 Chr. 1 - 1399575 - 1401270 [+] on Build GRCh38
          GenBankmRNA BX387394.2 - 3 718 Chr. 1 - 1399572 - 1401353 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0