Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC148413 SIRNA Assays ›
          Gene Symbol:
          LOC148413
          Gene Name:
          uncharacterized LOC148413
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 1399530 - 1402046 on Build GRCh38
          UniGene ID:
          Hs.741357
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n259660

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC148413
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015434.1 3 686
          GenBank mRNA AK024040.1 - 3 660
          AK092572.1 - 1 1179
          AK297832.1 - 3 675
          BC068600.1 - 3 643
          BC080546.1 - 3 642
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI767355.1 - 3 629 Chr. 1 - 1399582 - 1401172 [+] on Build GRCh38
          GenBankmRNA AL528291.3 - 3 816 Chr. 1 - 1399572 - 1401180 [+] on Build GRCh38
          GenBankmRNA BM921447.1 - 3 646 Chr. 1 - 1399572 - 1401278 [+] on Build GRCh38
          GenBankmRNA BI911209.1 - 3 725 Chr. 1 - 1399526 - 1401087 [+] on Build GRCh38
          GenBankmRNA BI758740.1 - 3 662 Chr. 1 - 1399550 - 1401296 [+] on Build GRCh38
          GenBankmRNA BX376671.2 - 3 663 Chr. 1 - 1399575 - 1401329 [+] on Build GRCh38
          GenBankmRNA BI517383.1 - 3 630 Chr. 1 - 1399581 - 1401219 [+] on Build GRCh38
          GenBankmRNA BQ058973.1 - 3 622 Chr. 1 - 1399594 - 1401350 [+] on Build GRCh38
          GenBankmRNA BE795124.1 - 619 Chr. - - [-] on Build
          GenBankmRNA BM547793.1 - 3 625 Chr. 1 - 1399591 - 1401261 [+] on Build GRCh38
          GenBankmRNA BQ064163.1 - 622 Chr. - - [-] on Build
          GenBankmRNA AL527639.3 - 3 639 Chr. 1 - 1399575 - 1401270 [+] on Build GRCh38
          GenBankmRNA DT220464.1 - 3 661 Chr. 1 - 1399550 - 1401000 [+] on Build GRCh38
          GenBankmRNA BX387394.2 - 3 638 Chr. 1 - 1399572 - 1401353 [+] on Build GRCh38
          GenBankmRNA BF037666.1 - 3 599 Chr. 1 - 1399612 - 1401215 [+] on Build GRCh38
          GenBankmRNA BE901076.1 - 3 633 Chr. 1 - 1399573 - 1401019 [+] on Build GRCh38
          GenBankmRNA BQ061816.1 - 3 623 Chr. 1 - 1399594 - 1401252 [+] on Build GRCh38
          GenBankmRNA AL548695.3 - 3 606 Chr. 1 - 1399606 - 1401277 [+] on Build GRCh38
          GenBankmRNA BI835920.1 - 3 642 Chr. 1 - 1399572 - 1401087 [+] on Build GRCh38
          GenBankmRNA BI257793.1 - 556 Chr. - - [-] on Build
          GenBankmRNA AL558971.3 - 3 643 Chr. 1 - 1399575 - 1401188 [+] on Build GRCh38
          GenBankmRNA AL533370.3 - 3 673 Chr. 1 - 1399561 - 1401295 [+] on Build GRCh38
          GenBankmRNA BQ069125.1 - 667 Chr. - - [-] on Build
          GenBankmRNA BI552758.1 - 3 649 Chr. 1 - 1399564 - 1401134 [+] on Build GRCh38
          GenBankmRNA CX754012.1 - 3 655 Chr. 1 - 1399555 - 1401063 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0