Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC148413 SIRNA Assays ›
          Gene Symbol:
          LOC148413
          Gene Name:
          uncharacterized LOC148413
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 1399530 - 1402046 on Build GRCh38
          UniGene ID:
          Hs.741357
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n259661

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC148413
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015434.1 3 534
          GenBank mRNA AK024040.1 - 3 508
          AK092572.1 - 1 1027
          AK297832.1 - 3 523
          BC068600.1 - 3 491
          BC080546.1 - 3 490
          BC104242.1 - 3 538
          BC104243.1 - 3 534
          BC112998.1 - 3 532
          BC112999.1 - 3 528
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI118146.1 - 3 476 Chr. 1 - 1399584 - 1401182 [+] on Build GRCh38
          GenBankmRNA CX758374.1 - 3 505 Chr. 1 - 1399557 - 1401019 [+] on Build GRCh38
          GenBankmRNA BX376671.2 - 3 513 Chr. 1 - 1399575 - 1401329 [+] on Build GRCh38
          GenBankmRNA DA698774.1 - 3 496 Chr. 1 - 1399564 - 1400871 [+] on Build GRCh38
          GenBankmRNA DA915106.1 - 3 482 Chr. 1 - 1399582 - 1400901 [+] on Build GRCh38
          GenBankmRNA BG254416.1 - 3 505 Chr. 1 - 1399555 - 1401146 [+] on Build GRCh38
          GenBankmRNA DA954863.1 - 3 480 Chr. 1 - 1399587 - 1400925 [+] on Build GRCh38
          GenBankmRNA BE795124.1 - 466 Chr. - - [-] on Build
          GenBankmRNA BQ064163.1 - 470 Chr. - - [-] on Build
          GenBankmRNA AU143273.1 - 3 508 Chr. 1 - 1399552 - 1401048 [+] on Build GRCh38
          GenBankmRNA DA356186.1 - 3 515 Chr. 1 - 1399552 - 1400880 [+] on Build GRCh38
          GenBankmRNA DA931499.1 - 3 477 Chr. 1 - 1399587 - 1400871 [+] on Build GRCh38
          GenBankmRNA AL527639.3 - 3 489 Chr. 1 - 1399575 - 1401270 [+] on Build GRCh38
          GenBankmRNA BX387394.2 - 3 486 Chr. 1 - 1399572 - 1401353 [+] on Build GRCh38
          GenBankmRNA DA489530.1 - 3 511 Chr. 1 - 1399548 - 1400868 [+] on Build GRCh38
          GenBankmRNA DC393491.1 - 3 512 Chr. 1 - 1399546 - 1400880 [+] on Build GRCh38
          GenBankmRNA BE901076.1 - 3 490 Chr. 1 - 1399573 - 1401019 [+] on Build GRCh38
          GenBankmRNA BQ061816.1 - 3 471 Chr. 1 - 1399594 - 1401252 [+] on Build GRCh38
          GenBankmRNA DA988281.1 - 3 506 Chr. 1 - 1399552 - 1400894 [+] on Build GRCh38
          GenBankmRNA AL558971.3 - 3 490 Chr. 1 - 1399575 - 1401188 [+] on Build GRCh38
          GenBankmRNA DA209785.1 - 3 512 Chr. 1 - 1399548 - 1400880 [+] on Build GRCh38
          GenBankmRNA BQ069125.1 - 515 Chr. - - [-] on Build
          GenBankmRNA BI552758.1 - 3 496 Chr. 1 - 1399564 - 1401134 [+] on Build GRCh38
          GenBankmRNA DB115625.1 - 3 499 Chr. 1 - 1399565 - 1400880 [+] on Build GRCh38
          GenBankmRNA BI767355.1 - 3 476 Chr. 1 - 1399582 - 1401172 [+] on Build GRCh38
          GenBankmRNA AL528291.3 - 3 665 Chr. 1 - 1399572 - 1401180 [+] on Build GRCh38
          GenBankmRNA BM921447.1 - 3 494 Chr. 1 - 1399572 - 1401278 [+] on Build GRCh38
          GenBankmRNA BI758740.1 - 3 510 Chr. 1 - 1399550 - 1401296 [+] on Build GRCh38
          GenBankmRNA BI911209.1 - 3 573 Chr. 1 - 1399526 - 1401087 [+] on Build GRCh38
          GenBankmRNA CR986326.1 - 3 484 Chr. 1 - 1399581 - 1400968 [+] on Build GRCh38
          GenBankmRNA BG328690.1 - 3 492 Chr. 1 - 1399573 - 1401103 [+] on Build GRCh38
          GenBankmRNA BI517383.1 - 3 479 Chr. 1 - 1399581 - 1401219 [+] on Build GRCh38
          GenBankmRNA BQ058973.1 - 3 470 Chr. 1 - 1399594 - 1401350 [+] on Build GRCh38
          GenBankmRNA DB262233.1 - 3 509 Chr. 1 - 1399552 - 1400902 [+] on Build GRCh38
          GenBankmRNA BM547793.1 - 3 473 Chr. 1 - 1399591 - 1401261 [+] on Build GRCh38
          GenBankmRNA DT220464.1 - 3 509 Chr. 1 - 1399550 - 1401000 [+] on Build GRCh38
          GenBankmRNA DA254561.1 - 3 508 Chr. 1 - 1399552 - 1400857 [+] on Build GRCh38
          GenBankmRNA DA895663.1 - 3 509 Chr. 1 - 1399552 - 1400872 [+] on Build GRCh38
          GenBankmRNA DA169042.1 - 3 461 Chr. 1 - 1399603 - 1400939 [+] on Build GRCh38
          GenBankmRNA BF037666.1 - 3 448 Chr. 1 - 1399612 - 1401215 [+] on Build GRCh38
          GenBankmRNA DC356520.1 - 3 523 Chr. 1 - 1399541 - 1400879 [+] on Build GRCh38
          GenBankmRNA CD580461.2 - 3 489 Chr. 1 - 1399581 - 1401143 [+] on Build GRCh38
          GenBankmRNA DA141713.1 - 3 497 Chr. 1 - 1399564 - 1400877 [+] on Build GRCh38
          GenBankmRNA DA132942.1 - 3 508 Chr. 1 - 1399552 - 1400872 [+] on Build GRCh38
          GenBankmRNA AL548695.3 - 3 456 Chr. 1 - 1399606 - 1401277 [+] on Build GRCh38
          GenBankmRNA BQ070201.1 - 3 488 Chr. 1 - 1399576 - 1401291 [+] on Build GRCh38
          GenBankmRNA BI835920.1 - 3 490 Chr. 1 - 1399572 - 1401087 [+] on Build GRCh38
          GenBankmRNA BI522617.1 - 3 531 Chr. 1 - 1399527 - 1400972 [+] on Build GRCh38
          GenBankmRNA DC381095.1 - 3 519 Chr. 1 - 1399541 - 1400857 [+] on Build GRCh38
          GenBankmRNA BI257793.1 - 405 Chr. - - [-] on Build
          GenBankmRNA DT218114.1 - 3 509 Chr. 1 - 1399550 - 1400923 [+] on Build GRCh38
          GenBankmRNA DA001807.1 - 3 500 Chr. 1 - 1399564 - 1400888 [+] on Build GRCh38
          GenBankmRNA AL533370.3 - 3 523 Chr. 1 - 1399561 - 1401295 [+] on Build GRCh38
          GenBankmRNA DB246546.1 - 3 515 Chr. 1 - 1399549 - 1400871 [+] on Build GRCh38
          GenBankmRNA DA061626.1 - 3 508 Chr. 1 - 1399552 - 1400898 [+] on Build GRCh38
          GenBankmRNA CX754012.1 - 3 503 Chr. 1 - 1399555 - 1401063 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0