Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RRN3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003370.2 | 12 | 1241 | |||
GenBank mRNA | BC006441.1 | - | 6 | 2240 | ||
BC112115.1 | - | 6 | 1369 | |||
BC112117.1 | - | 6 | 1369 |