Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RRP7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002184.2 | 7 | 1627 | |||
GenBank mRNA | BC007736.2 | - | 7 | 1627 |
Gene Symbol: RRP7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015703.4 | NP_056518.2 | 7 | 1659 | ||
GenBank mRNA | BC031838.1 | - | 6 | 1574 | ||
BC073834.1 | - | 7 | 1632 | 5533064 |