Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RRP7BP SIRNA Assays ›
          Gene Symbol:
          RRP7BP
          Gene Name:
          ribosomal RNA processing 7 homolog B, pseudogene
          Gene Aliases:
          RRP7B, dJ222E13.2
          Chromosome Location:
          Chr.22: 42573244 - 42582011 on Build GRCh38
          UniGene ID:
          Hs.534041
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n260089

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RRP7BP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002184.2 2 155
          GenBank mRNA AK098201.1 - 2 182
          AL157851.1 - 2 140
          BC007736.2 - 2 155
          BC014647.1 - 2 153
          BC143340.1 - 2 243
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI768330.1 - 2 148 Chr. 22 - 42574702 - 42582005 [-] on Build GRCh38
          GenBankmRNA DA010349.1 - 2 153 Chr. 22 - 42576523 - 42582009 [-] on Build GRCh38
          GenBankmRNA DB247769.1 - 2 183 Chr. 22 - 42576562 - 42582038 [-] on Build GRCh38
          GenBankmRNA CN418935.1 - 2 151 Chr. 22 - 42576052 - 42582007 [-] on Build GRCh38
          GenBankmRNA BP226541.1 - 2 153 Chr. 22 - 42574572 - 42582009 [-] on Build GRCh38
          GenBankmRNA BI917512.1 - 2 156 Chr. 22 - 42575956 - 42582012 [-] on Build GRCh38
          GenBankmRNA CN418933.1 - 2 144 Chr. 22 - 42576524 - 42581999 [-] on Build GRCh38
          GenBankmRNA DA264937.1 - 2 182 Chr. 22 - 42576562 - 42582038 [-] on Build GRCh38
          GenBankmRNA DA943625.1 - 2 163 Chr. 22 - 42576110 - 42582019 [-] on Build GRCh38
          GenBankmRNA CX754194.1 - 2 141 Chr. 22 - 42577096 - 42581997 [-] on Build GRCh38
          GenBankmRNA CD724112.1 - 2 156 Chr. 22 - 42576584 - 42582012 [-] on Build GRCh38
          GenBankmRNA BP202710.1 - 2 152 Chr. 22 - 42514282 - 42581999 [-] on Build GRCh38
          GenBankmRNA BX459908.2 - 2 187 Chr. 22 - 42556616 - 42582027 [-] on Build GRCh38
          GenBankmRNA BI830607.1 - 2 152 Chr. 22 - 42575948 - 42582009 [-] on Build GRCh38
          GenBankmRNA BM466918.1 - 165 Chr. - - [-] on Build
          GenBankmRNA BY799643.2 - 2 138 Chr. 22 - 42576066 - 42581994 [-] on Build GRCh38
          GenBankmRNA DA440686.1 - 2 182 Chr. 22 - 42576504 - 42582038 [-] on Build GRCh38
          GenBankmRNA DB457097.2 - 2 148 Chr. 22 - 42575979 - 42582004 [-] on Build GRCh38
          GenBankmRNA DA073621.1 - 2 156 Chr. 22 - 42576504 - 42582012 [-] on Build GRCh38
          GenBankmRNA CD246424.1 - 2 203 Chr. 22 - 42512911 - 42582036 [-] on Build GRCh38
          GenBankmRNA DA273984.1 - 2 182 Chr. 22 - 42576536 - 42582038 [-] on Build GRCh38
          GenBankmRNA DA919293.1 - 2 182 Chr. 22 - 42576532 - 42582035 [-] on Build GRCh38
          GenBankmRNA HY089361.1 - 2 180 Chr. 22 - 42576578 - 42582036 [-] on Build GRCh38
          GenBankmRNA BX339047.2 - 2 195 Chr. 22 - 42579165 - 42582035 [-] on Build GRCh38
          GenBankmRNA DA516063.1 - 2 181 Chr. 22 - 42576561 - 42582036 [-] on Build GRCh38
          GenBankmRNA BX446210.2 - 2 199 Chr. 22 - 42576512 - 42582027 [-] on Build GRCh38
          GenBankmRNA BX341133.2 - 2 145 Chr. 22 - 42574680 - 42582001 [-] on Build GRCh38
          GenBankmRNA DA259669.1 - 2 184 Chr. 22 - 42576570 - 42582040 [-] on Build GRCh38
          GenBankmRNA DA093686.1 - 2 151 Chr. 22 - 42576103 - 42582007 [-] on Build GRCh38
          GenBankmRNA BF304711.1 - 155 Chr. - - [-] on Build
          GenBankmRNA BI915045.1 - 2 156 Chr. 22 - 42514155 - 42582012 [-] on Build GRCh38
          GenBankmRNA BX446209.2 - 2 199 Chr. 22 - 42574704 - 42582027 [-] on Build GRCh38
          GenBankmRNA BG825249.1 - 2 156 Chr. 22 - 42514690 - 42582011 [-] on Build GRCh38
          GenBankmRNA DA819582.1 - 2 180 Chr. 22 - 42576537 - 42582036 [-] on Build GRCh38
          GenBankmRNA BG540871.1 - 2 156 Chr. 22 - 42576110 - 42582011 [-] on Build GRCh38
          GenBankmRNA DA938775.1 - 2 155 Chr. 22 - 42576570 - 42582011 [-] on Build GRCh38
          GenBankmRNA DA924704.1 - 2 182 Chr. 22 - 42576527 - 42582038 [-] on Build GRCh38
          GenBankmRNA DA358331.1 - 2 182 Chr. 22 - 42576544 - 42582038 [-] on Build GRCh38
          GenBankmRNA AL603051.1 - 2 153 Chr. 22 - 42576052 - 42582009 [-] on Build GRCh38
          GenBankmRNA BJ993265.1 - 2 155 Chr. 22 - 42576560 - 42582011 [-] on Build GRCh38
          GenBankmRNA BP196344.1 - 2 150 Chr. 22 - 42576511 - 42582006 [-] on Build GRCh38
          GenBankmRNA CR747287.1 - 2 155 Chr. 22 - 42576574 - 42582011 [-] on Build GRCh38
          GenBankmRNA BM545074.1 - 2 185 Chr. 22 - 42556561 - 42582022 [-] on Build GRCh38
          GenBankmRNA DA932993.1 - 2 153 Chr. 22 - 42576507 - 42582009 [-] on Build GRCh38
          GenBankmRNA BG470664.1 - 2 154 Chr. 22 - 42576077 - 42582010 [-] on Build GRCh38
          GenBankmRNA DA485392.1 - 2 151 Chr. 22 - 42576522 - 42582007 [-] on Build GRCh38
          GenBankmRNA DA947432.1 - 2 182 Chr. 22 - 42576508 - 42582038 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s)

          ribosomal small subunit assembly
          rRNA processing

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0