Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RRP7BP SIRNA Assays ›
          Gene Symbol:
          RRP7BP
          Gene Name:
          ribosomal RNA processing 7 homolog B, pseudogene
          Gene Aliases:
          RRP7B, dJ222E13.2
          Chromosome Location:
          Chr.22: 42573244 - 42582011 on Build GRCh38
          UniGene ID:
          Hs.534041
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n260093

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RRP7BP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002184.2 2 198
          GenBank mRNA AK098201.1 - 2 225
          AL157851.1 - 2 183
          BC007736.2 - 2 198
          BC014647.1 - 2 196
          BC143340.1 - 2 286
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA023427.1 - 2 225 Chr. 22 - 42576536 - 42582038 [-] on Build GRCh38
          GenBankmRNA DB048618.1 - 2 197 Chr. 22 - 42576093 - 42582009 [-] on Build GRCh38
          GenBankmRNA DA010349.1 - 2 196 Chr. 22 - 42576523 - 42582009 [-] on Build GRCh38
          GenBankmRNA DB247769.1 - 2 226 Chr. 22 - 42576562 - 42582038 [-] on Build GRCh38
          GenBankmRNA DA868852.1 - 2 197 Chr. 22 - 42576545 - 42582009 [-] on Build GRCh38
          GenBankmRNA DA082677.1 - 2 217 Chr. 22 - 42576514 - 42582029 [-] on Build GRCh38
          GenBankmRNA DA778313.1 - 2 213 Chr. 22 - 42576522 - 42582025 [-] on Build GRCh38
          GenBankmRNA CN418935.1 - 2 194 Chr. 22 - 42576052 - 42582007 [-] on Build GRCh38
          GenBankmRNA BP226541.1 - 2 196 Chr. 22 - 42574572 - 42582009 [-] on Build GRCh38
          GenBankmRNA CN418933.1 - 2 187 Chr. 22 - 42576524 - 42581999 [-] on Build GRCh38
          GenBankmRNA DA264937.1 - 2 225 Chr. 22 - 42576562 - 42582038 [-] on Build GRCh38
          GenBankmRNA DA240322.1 - 2 209 Chr. 22 - 42576103 - 42582019 [-] on Build GRCh38
          GenBankmRNA CX754194.1 - 2 184 Chr. 22 - 42577096 - 42581997 [-] on Build GRCh38
          GenBankmRNA CD724112.1 - 2 199 Chr. 22 - 42576584 - 42582012 [-] on Build GRCh38
          GenBankmRNA DA943625.1 - 2 206 Chr. 22 - 42576110 - 42582019 [-] on Build GRCh38
          GenBankmRNA BP202710.1 - 2 195 Chr. 22 - 42514282 - 42581999 [-] on Build GRCh38
          GenBankmRNA BX459908.2 - 2 230 Chr. 22 - 42556616 - 42582027 [-] on Build GRCh38
          GenBankmRNA DA106662.1 - 2 225 Chr. 22 - 42576545 - 42582038 [-] on Build GRCh38
          GenBankmRNA BI830607.1 - 2 195 Chr. 22 - 42575948 - 42582009 [-] on Build GRCh38
          GenBankmRNA BM466918.1 - 208 Chr. - - [-] on Build
          GenBankmRNA DB048644.1 - 2 197 Chr. 22 - 42576095 - 42582009 [-] on Build GRCh38
          GenBankmRNA DB108619.1 - 2 205 Chr. 22 - 42576108 - 42582017 [-] on Build GRCh38
          GenBankmRNA BY799643.2 - 2 181 Chr. 22 - 42576066 - 42581994 [-] on Build GRCh38
          GenBankmRNA DB127475.1 - 2 214 Chr. 22 - 42576534 - 42582026 [-] on Build GRCh38
          GenBankmRNA DB457097.2 - 2 191 Chr. 22 - 42575979 - 42582004 [-] on Build GRCh38
          GenBankmRNA DA073621.1 - 2 199 Chr. 22 - 42576504 - 42582012 [-] on Build GRCh38
          GenBankmRNA DA273984.1 - 2 225 Chr. 22 - 42576536 - 42582038 [-] on Build GRCh38
          GenBankmRNA CD246424.1 - 2 246 Chr. 22 - 42512911 - 42582036 [-] on Build GRCh38
          GenBankmRNA DA944802.1 - 2 197 Chr. 22 - 42576535 - 42582009 [-] on Build GRCh38
          GenBankmRNA DA042801.1 - 2 205 Chr. 22 - 42576504 - 42582017 [-] on Build GRCh38
          GenBankmRNA DB056697.1 - 2 195 Chr. 22 - 42576981 - 42582007 [-] on Build GRCh38
          GenBankmRNA DA919293.1 - 2 225 Chr. 22 - 42576532 - 42582035 [-] on Build GRCh38
          GenBankmRNA HY089361.1 - 2 223 Chr. 22 - 42576578 - 42582036 [-] on Build GRCh38
          GenBankmRNA BX339047.2 - 2 238 Chr. 22 - 42579165 - 42582035 [-] on Build GRCh38
          GenBankmRNA BX446210.2 - 2 242 Chr. 22 - 42576512 - 42582027 [-] on Build GRCh38
          GenBankmRNA DA516063.1 - 2 224 Chr. 22 - 42576561 - 42582036 [-] on Build GRCh38
          GenBankmRNA DA566262.1 - 2 205 Chr. 22 - 42576512 - 42582017 [-] on Build GRCh38
          GenBankmRNA DA259669.1 - 2 227 Chr. 22 - 42576570 - 42582040 [-] on Build GRCh38
          GenBankmRNA DA093686.1 - 2 194 Chr. 22 - 42576103 - 42582007 [-] on Build GRCh38
          GenBankmRNA BX341133.2 - 2 188 Chr. 22 - 42574680 - 42582001 [-] on Build GRCh38
          GenBankmRNA DB192376.1 - 2 214 Chr. 22 - 42576532 - 42582026 [-] on Build GRCh38
          GenBankmRNA BX446209.2 - 2 242 Chr. 22 - 42574704 - 42582027 [-] on Build GRCh38
          GenBankmRNA BI915045.1 - 2 199 Chr. 22 - 42514155 - 42582012 [-] on Build GRCh38
          GenBankmRNA DA138190.1 - 2 209 Chr. 22 - 42576533 - 42582021 [-] on Build GRCh38
          GenBankmRNA DA156810.1 - 2 193 Chr. 22 - 42576504 - 42582005 [-] on Build GRCh38
          GenBankmRNA BG825249.1 - 2 199 Chr. 22 - 42514690 - 42582011 [-] on Build GRCh38
          GenBankmRNA DA819582.1 - 2 223 Chr. 22 - 42576537 - 42582036 [-] on Build GRCh38
          GenBankmRNA BG540871.1 - 2 199 Chr. 22 - 42576110 - 42582011 [-] on Build GRCh38
          GenBankmRNA DA938775.1 - 2 198 Chr. 22 - 42576570 - 42582011 [-] on Build GRCh38
          GenBankmRNA DA939725.1 - 2 198 Chr. 22 - 42576511 - 42582011 [-] on Build GRCh38
          GenBankmRNA DA209120.1 - 2 197 Chr. 22 - 42576986 - 42582009 [-] on Build GRCh38
          GenBankmRNA DA924704.1 - 2 225 Chr. 22 - 42576527 - 42582038 [-] on Build GRCh38
          GenBankmRNA BM805010.1 - 262 Chr. - - [-] on Build
          GenBankmRNA DA880214.1 - 2 226 Chr. 22 - 42576543 - 42582038 [-] on Build GRCh38
          GenBankmRNA DA521095.1 - 2 194 Chr. 22 - 42576511 - 42582006 [-] on Build GRCh38
          GenBankmRNA DA358331.1 - 2 225 Chr. 22 - 42576544 - 42582038 [-] on Build GRCh38
          GenBankmRNA AL603051.1 - 2 196 Chr. 22 - 42576052 - 42582009 [-] on Build GRCh38
          GenBankmRNA BP196344.1 - 2 193 Chr. 22 - 42576511 - 42582006 [-] on Build GRCh38
          GenBankmRNA BJ993265.1 - 2 198 Chr. 22 - 42576560 - 42582011 [-] on Build GRCh38
          GenBankmRNA DA265731.1 - 2 214 Chr. 22 - 42514692 - 42582026 [-] on Build GRCh38
          GenBankmRNA CR747287.1 - 2 198 Chr. 22 - 42576574 - 42582011 [-] on Build GRCh38
          GenBankmRNA BM545074.1 - 2 228 Chr. 22 - 42556561 - 42582022 [-] on Build GRCh38
          GenBankmRNA DB172296.1 - 2 226 Chr. 22 - 42576520 - 42582038 [-] on Build GRCh38
          GenBankmRNA DA932993.1 - 2 196 Chr. 22 - 42576507 - 42582009 [-] on Build GRCh38
          GenBankmRNA BG470664.1 - 2 197 Chr. 22 - 42576077 - 42582010 [-] on Build GRCh38
          GenBankmRNA DA485392.1 - 2 194 Chr. 22 - 42576522 - 42582007 [-] on Build GRCh38
          GenBankmRNA DA947432.1 - 2 225 Chr. 22 - 42576508 - 42582038 [-] on Build GRCh38
          GenBankmRNA DA070601.1 - 2 209 Chr. 22 - 42576525 - 42582020 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s)

          ribosomal small subunit assembly
          rRNA processing

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0