Search
Search

LINC00889| Gene Symbol: LINC00889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026935.1 | 1 | 1734 | |||
| GenBank mRNA | AK055234.1 | - | 1 | 1283 | ||
| AK055382.1 | - | 1 | 1418 | |||
| BC105719.1 | - | 1 | 1408 | |||
| BC105725.1 | - | 1 | 1408 | |||
| BC105726.1 | - | 1 | 1408 | |||
| BC105727.1 | - | 1 | 1408 | |||
| BC139831.1 | - | 1 | 1411 | |||