Search
Search

LINC00889| Gene Symbol: LINC00889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026935.1 | 1 | 2008 | |||
| GenBank mRNA | AK055234.1 | - | 1 | 1557 | ||
| AK055382.1 | - | 1 | 1692 | |||
| BC105719.1 | - | 1 | 1682 | |||
| BC105725.1 | - | 1 | 1682 | |||
| BC105726.1 | - | 1 | 1682 | |||
| BC105727.1 | - | 1 | 1682 | |||
| BC139831.1 | - | 1 | 1685 | |||