Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMEM79 SIRNA Assays ›
          Gene Symbol:
          TMEM79
          Gene Name:
          transmembrane protein 79
          Gene Aliases:
          MATT
          Chromosome Location:
          Chr.1: 156282913 - 156292443 on Build GRCh38
          UniGene ID:
          Hs.347408
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n261582

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMEM79
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026678.1 1 73
          GenBank mRNA AK122634.1 - 1 51
          BC005094.1 - 1 38 3955668
          DA678865.1 - 1 73
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA055657.1 - 1 59 Chr. 1 - 156282927 - 156285595 [+] on Build GRCh38
          GenBankmRNA CB141162.1 - 1 51 Chr. 1 - 156282935 - 156285667 [+] on Build GRCh38
          GenBankmRNA DA746201.1 - 1 52 Chr. 1 - 156282934 - 156285605 [+] on Build GRCh38
          GenBankmRNA DA341511.1 - 1 49 Chr. 1 - 156282937 - 156285613 [+] on Build GRCh38
          GenBankmRNA DA936318.1 - 1 51 Chr. 1 - 156282935 - 156285655 [+] on Build GRCh38
          GenBankmRNA DA385665.1 - 1 57 Chr. 1 - 156282929 - 156285635 [+] on Build GRCh38
          GenBankmRNA DA002835.1 - 1 51 Chr. 1 - 156282935 - 156285619 [+] on Build GRCh38
          GenBankmRNA DA867329.1 - 1 51 Chr. 1 - 156282935 - 156285602 [+] on Build GRCh38
          GenBankmRNA AA595617.1 - 1 20 Chr. 1 - 156282966 - 156291689 [+] on Build GRCh38
          GenBankmRNA DA900776.1 - 1 74 Chr. 1 - 156282912 - 156285755 [+] on Build GRCh38
          GenBankmRNA DA706610.1 - 1 32 Chr. 1 - 156282954 - 156285600 [+] on Build GRCh38
          GenBankmRNA DA553808.1 - 1 51 Chr. 1 - 156282935 - 156285617 [+] on Build GRCh38
          GenBankmRNA DA433954.1 - 1 49 Chr. 1 - 156282937 - 156285581 [+] on Build GRCh38
          GenBankmRNA DA955071.1 - 1 51 Chr. 1 - 156282935 - 156285623 [+] on Build GRCh38
          GenBankmRNA DB116598.1 - 1 51 Chr. 1 - 156282935 - 156285592 [+] on Build GRCh38
          GenBankmRNA BG104443.1 - 1 39 Chr. 1 - 156282947 - 156285740 [+] on Build GRCh38
          GenBankmRNA DA044506.1 - 1 51 Chr. 1 - 156282935 - 156285612 [+] on Build GRCh38
          GenBankmRNA DA287771.1 - 1 57 Chr. 1 - 156282929 - 156285580 [+] on Build GRCh38
          GenBankmRNA DA629602.1 - 1 37 Chr. 1 - 156282952 - 156285639 [+] on Build GRCh38
          GenBankmRNA DA866716.1 - 1 59 Chr. 1 - 156282927 - 156285586 [+] on Build GRCh38
          GenBankmRNA DA949224.1 - 1 51 Chr. 1 - 156282935 - 156285625 [+] on Build GRCh38
          GenBankmRNA DA934185.1 - 1 67 Chr. 1 - 156282919 - 156285612 [+] on Build GRCh38
          GenBankmRNA HY069344.1 - 1 61 Chr. 1 - 156282925 - 156285517 [+] on Build GRCh38
          GenBankmRNA DA205113.1 - 1 51 Chr. 1 - 156282935 - 156285614 [+] on Build GRCh38
          GenBankmRNA DA676249.1 - 1 45 Chr. 1 - 156282941 - 156285649 [+] on Build GRCh38
          GenBankmRNA DN997286.1 - 1 31 Chr. 1 - 156282963 - 156285754 [+] on Build GRCh38
          GenBankmRNA DA868906.1 - 1 35 Chr. 1 - 156282951 - 156285604 [+] on Build GRCh38
          GenBankmRNA DA494865.1 - 1 59 Chr. 1 - 156282927 - 156285580 [+] on Build GRCh38
          GenBankmRNA AL559952.3 - 1 50 Chr. 1 - 156282936 - 156286407 [+] on Build GRCh38
          GenBankmRNA DA347460.1 - 1 59 Chr. 1 - 156282927 - 156285580 [+] on Build GRCh38
          GenBankmRNA DB474483.1 - 1 51 Chr. 1 - 156282935 - 156285537 [+] on Build GRCh38
          GenBankmRNA BI821654.1 - 1 56 Chr. 1 - 156282931 - 156285785 [+] on Build GRCh38
          GenBankmRNA DB107978.1 - 1 35 Chr. 1 - 156282951 - 156285637 [+] on Build GRCh38
          GenBankmRNA HY071423.1 - 1 58 Chr. 1 - 156282929 - 156285515 [+] on Build GRCh38
          GenBankmRNA DA879928.1 - 1 51 Chr. 1 - 156282935 - 156285634 [+] on Build GRCh38
          GenBankmRNA AA579343.1 - 1 22 Chr. 1 - 156282964 - 156285317 [+] on Build GRCh38
          GenBankmRNA DB009066.1 - 1 51 Chr. 1 - 156282935 - 156285863 [+] on Build GRCh38
          GenBankmRNA DB115121.1 - 1 52 Chr. 1 - 156282934 - 156285605 [+] on Build GRCh38
          GenBankmRNA DA437863.1 - 1 51 Chr. 1 - 156282935 - 156285619 [+] on Build GRCh38
          GenBankmRNA DA344041.1 - 1 51 Chr. 1 - 156282935 - 156285638 [+] on Build GRCh38
          GenBankmRNA DB118285.1 - 1 51 Chr. 1 - 156282935 - 156285606 [+] on Build GRCh38
          GenBankmRNA DA377167.1 - 1 58 Chr. 1 - 156282929 - 156285597 [+] on Build GRCh38
          GenBankmRNA DA848063.1 - 1 52 Chr. 1 - 156282935 - 156285612 [+] on Build GRCh38
          GenBankmRNA DA831678.1 - 1 51 Chr. 1 - 156282935 - 156285804 [+] on Build GRCh38
          GenBankmRNA DA344165.1 - 1 59 Chr. 1 - 156282927 - 156285622 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial cell maturation
          hair follicle morphogenesis
          cuticle development
          regulated exocytosis
          positive regulation of epidermis development
          establishment of skin barrier
          cornification
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0