Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMEM79 SIRNA Assays ›
          Gene Symbol:
          TMEM79
          Gene Name:
          transmembrane protein 79
          Gene Aliases:
          MATT
          Chromosome Location:
          Chr.1: 156282913 - 156292443 on Build GRCh38
          UniGene ID:
          Hs.347408
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n261586

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMEM79
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026678.1 1 128
          GenBank mRNA AK122634.1 - 1 106
          BC005094.1 - 1 93 3955668
          DA678865.1 - 1 128
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA HY110076.1 - 1 32 Chr. 1 - 156283009 - 156285662 [+] on Build GRCh38
          GenBankmRNA DA055657.1 - 1 114 Chr. 1 - 156282927 - 156285595 [+] on Build GRCh38
          GenBankmRNA CB141162.1 - 1 106 Chr. 1 - 156282935 - 156285667 [+] on Build GRCh38
          GenBankmRNA DA746201.1 - 1 107 Chr. 1 - 156282934 - 156285605 [+] on Build GRCh38
          GenBankmRNA DA341511.1 - 1 104 Chr. 1 - 156282937 - 156285613 [+] on Build GRCh38
          GenBankmRNA DA744821.1 - 1 30 Chr. 1 - 156283011 - 156285679 [+] on Build GRCh38
          GenBankmRNA DA385665.1 - 1 112 Chr. 1 - 156282929 - 156285635 [+] on Build GRCh38
          GenBankmRNA DA936318.1 - 1 106 Chr. 1 - 156282935 - 156285655 [+] on Build GRCh38
          GenBankmRNA DA002835.1 - 1 106 Chr. 1 - 156282935 - 156285619 [+] on Build GRCh38
          GenBankmRNA DA172530.1 - 1 34 Chr. 1 - 156283011 - 156285691 [+] on Build GRCh38
          GenBankmRNA DA867329.1 - 1 106 Chr. 1 - 156282935 - 156285602 [+] on Build GRCh38
          GenBankmRNA AA595617.1 - 1 75 Chr. 1 - 156282966 - 156291689 [+] on Build GRCh38
          GenBankmRNA DA900776.1 - 1 129 Chr. 1 - 156282912 - 156285755 [+] on Build GRCh38
          GenBankmRNA BI335549.1 - 1 118 Chr. 1 - 156282963 - 156285621 [+] on Build GRCh38
          GenBankmRNA DA706610.1 - 1 87 Chr. 1 - 156282954 - 156285600 [+] on Build GRCh38
          GenBankmRNA DA553808.1 - 1 106 Chr. 1 - 156282935 - 156285617 [+] on Build GRCh38
          GenBankmRNA DA433954.1 - 1 104 Chr. 1 - 156282937 - 156285581 [+] on Build GRCh38
          GenBankmRNA DA353123.1 - 1 27 Chr. 1 - 156283014 - 156285684 [+] on Build GRCh38
          GenBankmRNA DA955071.1 - 1 106 Chr. 1 - 156282935 - 156285623 [+] on Build GRCh38
          GenBankmRNA DA871821.1 - 1 54 Chr. 1 - 156282987 - 156285650 [+] on Build GRCh38
          GenBankmRNA DB116598.1 - 1 106 Chr. 1 - 156282935 - 156285592 [+] on Build GRCh38
          GenBankmRNA DA353342.1 - 1 33 Chr. 1 - 156283008 - 156285704 [+] on Build GRCh38
          GenBankmRNA BM474421.1 - 1 87 Chr. 1 - 156282953 - 156291907 [+] on Build GRCh38
          GenBankmRNA AU280322.1 - 1 55 Chr. 1 - 156282987 - 156285663 [+] on Build GRCh38
          GenBankmRNA BG104443.1 - 1 94 Chr. 1 - 156282947 - 156285740 [+] on Build GRCh38
          GenBankmRNA DA044506.1 - 1 106 Chr. 1 - 156282935 - 156285612 [+] on Build GRCh38
          GenBankmRNA DA287771.1 - 1 112 Chr. 1 - 156282929 - 156285580 [+] on Build GRCh38
          GenBankmRNA DA555582.1 - 1 56 Chr. 1 - 156282987 - 156285675 [+] on Build GRCh38
          GenBankmRNA BG744376.1 - 1 38 Chr. 1 - 156283004 - 156285785 [+] on Build GRCh38
          GenBankmRNA DA913069.1 - 1 33 Chr. 1 - 156283008 - 156285684 [+] on Build GRCh38
          GenBankmRNA DA625594.1 - 1 33 Chr. 1 - 156283008 - 156285672 [+] on Build GRCh38
          GenBankmRNA DA866716.1 - 1 114 Chr. 1 - 156282927 - 156285586 [+] on Build GRCh38
          GenBankmRNA DA949224.1 - 1 106 Chr. 1 - 156282935 - 156285625 [+] on Build GRCh38
          GenBankmRNA HY069344.1 - 1 116 Chr. 1 - 156282925 - 156285517 [+] on Build GRCh38
          GenBankmRNA DA934185.1 - 1 122 Chr. 1 - 156282919 - 156285612 [+] on Build GRCh38
          GenBankmRNA DA205113.1 - 1 106 Chr. 1 - 156282935 - 156285614 [+] on Build GRCh38
          GenBankmRNA DA676249.1 - 1 100 Chr. 1 - 156282941 - 156285649 [+] on Build GRCh38
          GenBankmRNA DN997286.1 - 1 86 Chr. 1 - 156282963 - 156285754 [+] on Build GRCh38
          GenBankmRNA DA868906.1 - 1 90 Chr. 1 - 156282951 - 156285604 [+] on Build GRCh38
          GenBankmRNA DA494865.1 - 1 114 Chr. 1 - 156282927 - 156285580 [+] on Build GRCh38
          GenBankmRNA AL559952.3 - 1 105 Chr. 1 - 156282936 - 156286407 [+] on Build GRCh38
          GenBankmRNA DA347460.1 - 1 114 Chr. 1 - 156282927 - 156285580 [+] on Build GRCh38
          GenBankmRNA DA697386.1 - 1 45 Chr. 1 - 156282996 - 156285677 [+] on Build GRCh38
          GenBankmRNA DB474483.1 - 1 106 Chr. 1 - 156282935 - 156285537 [+] on Build GRCh38
          GenBankmRNA DB107978.1 - 1 90 Chr. 1 - 156282951 - 156285637 [+] on Build GRCh38
          GenBankmRNA BI821654.1 - 1 111 Chr. 1 - 156282931 - 156285785 [+] on Build GRCh38
          GenBankmRNA HY071423.1 - 1 113 Chr. 1 - 156282929 - 156285515 [+] on Build GRCh38
          GenBankmRNA DA879928.1 - 1 106 Chr. 1 - 156282935 - 156285634 [+] on Build GRCh38
          GenBankmRNA DA639244.1 - 1 29 Chr. 1 - 156283012 - 156285685 [+] on Build GRCh38
          GenBankmRNA AA579343.1 - 1 77 Chr. 1 - 156282964 - 156285317 [+] on Build GRCh38
          GenBankmRNA DB009066.1 - 1 106 Chr. 1 - 156282935 - 156285863 [+] on Build GRCh38
          GenBankmRNA DB115121.1 - 1 107 Chr. 1 - 156282934 - 156285605 [+] on Build GRCh38
          GenBankmRNA DA344041.1 - 1 106 Chr. 1 - 156282935 - 156285638 [+] on Build GRCh38
          GenBankmRNA DA437863.1 - 1 106 Chr. 1 - 156282935 - 156285619 [+] on Build GRCh38
          GenBankmRNA DB047930.1 - 1 33 Chr. 1 - 156283009 - 156285680 [+] on Build GRCh38
          GenBankmRNA DB118285.1 - 1 106 Chr. 1 - 156282935 - 156285606 [+] on Build GRCh38
          GenBankmRNA DA377167.1 - 1 113 Chr. 1 - 156282929 - 156285597 [+] on Build GRCh38
          GenBankmRNA DA157348.1 - 1 40 Chr. 1 - 156283002 - 156285675 [+] on Build GRCh38
          GenBankmRNA DA848063.1 - 1 107 Chr. 1 - 156282935 - 156285612 [+] on Build GRCh38
          GenBankmRNA DA831678.1 - 1 106 Chr. 1 - 156282935 - 156285804 [+] on Build GRCh38
          GenBankmRNA DA344165.1 - 1 114 Chr. 1 - 156282927 - 156285622 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial cell maturation
          hair follicle morphogenesis
          cuticle development
          regulated exocytosis
          positive regulation of epidermis development
          establishment of skin barrier
          cornification
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0