Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RFPL3S SIRNA Assays ›
          Gene Symbol:
          RFPL3S
          Gene Name:
          RFPL3 antisense
          Gene Aliases:
          NCRNA00005, RFPL3-AS, RFPL3-AS1, RFPL3ANT, RFPL3AS
          Chromosome Location:
          Chr.22: 32359906 - 32371264 on Build GRCh38
          UniGene ID:
          Hs.274285
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n261854

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RFPL3S
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_001450.3 1 167
          NR_002596.2 1 167
          NR_104232.1 2 99
          GenBank mRNA AJ010233.1 - 1 76
          BC042824.1 - 1 93
          BC066983.1 - 1 93
          BC090926.1 - 1 93
          HY013491.1 - 2 99
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA HY028065.1 - 1 94 Chr. 22 - 32360641 - 32371002 [-] on Build GRCh38
          GenBankmRNA BI828584.1 - 1 167 Chr. 22 - 32360538 - 32371076 [-] on Build GRCh38
          GenBankmRNA HY040615.1 - 1 94 Chr. 22 - 32360579 - 32371003 [-] on Build GRCh38
          GenBankmRNA HY010615.1 - 1 94 Chr. 22 - 32360508 - 32371002 [-] on Build GRCh38
          GenBankmRNA DB457022.1 - 1 94 Chr. 22 - 32360628 - 32371003 [-] on Build GRCh38
          GenBankmRNA HY033409.1 - 1 94 Chr. 22 - 32360680 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY035024.1 - 1 123 Chr. 22 - 32360541 - 32371031 [-] on Build GRCh38
          GenBankmRNA BG185333.1 - 2 139 Chr. 22 - 32360761 - 32381253 [-] on Build GRCh38
          GenBankmRNA DB459325.1 - 1 90 Chr. 22 - 32360647 - 32370999 [-] on Build GRCh38
          GenBankmRNA HY030434.1 - 1 94 Chr. 22 - 32360589 - 32371003 [-] on Build GRCh38
          GenBankmRNA BG198113.1 - 2 138 Chr. 22 - 32360219 - 32381253 [-] on Build GRCh38
          GenBankmRNA HY026769.1 - 1 156 Chr. 22 - 32360701 - 32371064 [-] on Build GRCh38
          GenBankmRNA DB448776.1 - 1 139 Chr. 22 - 32360738 - 32371047 [-] on Build GRCh38
          GenBankmRNA HY049960.1 - 1 94 Chr. 22 - 32360692 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY017315.1 - 1 94 Chr. 22 - 32360581 - 32371002 [-] on Build GRCh38
          GenBankmRNA AA393375.1 - 1 76 Chr. 22 - 32360488 - 32370985 [-] on Build GRCh38
          GenBankmRNA DB455123.1 - 1 94 Chr. 22 - 32360647 - 32371003 [-] on Build GRCh38
          GenBankmRNA HY027374.1 - 1 94 Chr. 22 - 32360612 - 32371002 [-] on Build GRCh38
          GenBankmRNA BX117543.1 - 1 76 Chr. 22 - 32360271 - 32370985 [-] on Build GRCh38
          GenBankmRNA DB449158.1 - 1 162 Chr. 22 - 32360775 - 32371070 [-] on Build GRCh38
          GenBankmRNA HY027331.1 - 1 94 Chr. 22 - 32360615 - 32371002 [-] on Build GRCh38
          GenBankmRNA CN411545.1 - 4 297 Chr. 22 - 32362974 - 32382052 [-] on Build GRCh38
          GenBankmRNA BU584156.1 - 1 20 Chr. 22 - 32362977 - 32370929 [-] on Build GRCh38
          GenBankmRNA BU569141.1 - 1 96 Chr. 22 - 32360388 - 32371002 [-] on Build GRCh38
          GenBankmRNA BG723497.1 - 1 98 Chr. 22 - 32360422 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY015298.1 - 1 98 Chr. 22 - 32360592 - 32371004 [-] on Build GRCh38
          GenBankmRNA BU584154.1 - 1 20 Chr. 22 - 32363015 - 32370929 [-] on Build GRCh38
          GenBankmRNA CD359353.1 - 1 79 Chr. 22 - 32360312 - 32370990 [-] on Build GRCh38
          GenBankmRNA DB457839.1 - 1 90 Chr. 22 - 32360630 - 32370999 [-] on Build GRCh38
          GenBankmRNA DB447067.1 - 1 90 Chr. 22 - 32360698 - 32370999 [-] on Build GRCh38
          GenBankmRNA BI461189.1 - 1 98 Chr. 22 - 32360588 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY009642.1 - 1 94 Chr. 22 - 32360581 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY028203.1 - 1 94 Chr. 22 - 32360565 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY004848.1 - 1 94 Chr. 22 - 32360571 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY054907.1 - 1 94 Chr. 22 - 32360703 - 32371002 [-] on Build GRCh38
          GenBankmRNA BG771936.1 - 1 98 Chr. 22 - 32360389 - 32371002 [-] on Build GRCh38
          GenBankmRNA BG720986.1 - 1 98 Chr. 22 - 32360230 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY034267.1 - 1 94 Chr. 22 - 32360596 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY011074.1 - 1 94 Chr. 22 - 32360626 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY007602.1 - 1 139 Chr. 22 - 32360684 - 32371047 [-] on Build GRCh38
          GenBankmRNA HY002386.1 - 1 94 Chr. 22 - 32360553 - 32371003 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0