Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RFPL3S SIRNA Assays ›
          Gene Symbol:
          RFPL3S
          Gene Name:
          RFPL3 antisense
          Gene Aliases:
          NCRNA00005, RFPL3-AS, RFPL3-AS1, RFPL3ANT, RFPL3AS
          Chromosome Location:
          Chr.22: 32359906 - 32371264 on Build GRCh38
          UniGene ID:
          Hs.274285
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n261861

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RFPL3S
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_001450.3 2 205
          NR_002596.2 2 205
          NR_104232.1 3 137
          GenBank mRNA AJ010233.1 - 2 114
          BC042824.1 - 2 131
          BC066983.1 - 2 131
          BC090926.1 - 2 131
          HY013491.1 - 3 137
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA HY028065.1 - 2 132 Chr. 22 - 32360641 - 32371002 [-] on Build GRCh38
          GenBankmRNA BI828584.1 - 2 205 Chr. 22 - 32360538 - 32371076 [-] on Build GRCh38
          GenBankmRNA HY010615.1 - 2 132 Chr. 22 - 32360508 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY040615.1 - 2 132 Chr. 22 - 32360579 - 32371003 [-] on Build GRCh38
          GenBankmRNA DB442643.1 - 1 29 Chr. 22 - 32360564 - 32368569 [-] on Build GRCh38
          GenBankmRNA DB457022.1 - 2 132 Chr. 22 - 32360628 - 32371003 [-] on Build GRCh38
          GenBankmRNA HY033409.1 - 2 132 Chr. 22 - 32360680 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY035024.1 - 2 161 Chr. 22 - 32360541 - 32371031 [-] on Build GRCh38
          GenBankmRNA BG185333.1 - 3 177 Chr. 22 - 32360761 - 32381253 [-] on Build GRCh38
          GenBankmRNA HY030434.1 - 2 132 Chr. 22 - 32360589 - 32371003 [-] on Build GRCh38
          GenBankmRNA DB459325.1 - 2 128 Chr. 22 - 32360647 - 32370999 [-] on Build GRCh38
          GenBankmRNA BG198113.1 - 3 176 Chr. 22 - 32360219 - 32381253 [-] on Build GRCh38
          GenBankmRNA HY026769.1 - 2 194 Chr. 22 - 32360701 - 32371064 [-] on Build GRCh38
          GenBankmRNA DB448776.1 - 2 177 Chr. 22 - 32360738 - 32371047 [-] on Build GRCh38
          GenBankmRNA HY049960.1 - 2 132 Chr. 22 - 32360692 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY017315.1 - 2 132 Chr. 22 - 32360581 - 32371002 [-] on Build GRCh38
          GenBankmRNA AA393375.1 - 2 114 Chr. 22 - 32360488 - 32370985 [-] on Build GRCh38
          GenBankmRNA DB455123.1 - 2 132 Chr. 22 - 32360647 - 32371003 [-] on Build GRCh38
          GenBankmRNA HY027374.1 - 2 132 Chr. 22 - 32360612 - 32371002 [-] on Build GRCh38
          GenBankmRNA BX117543.1 - 2 114 Chr. 22 - 32360271 - 32370985 [-] on Build GRCh38
          GenBankmRNA DB449158.1 - 2 200 Chr. 22 - 32360775 - 32371070 [-] on Build GRCh38
          GenBankmRNA HY027331.1 - 2 132 Chr. 22 - 32360615 - 32371002 [-] on Build GRCh38
          GenBankmRNA CN411545.1 - 5 335 Chr. 22 - 32362974 - 32382052 [-] on Build GRCh38
          GenBankmRNA BU584156.1 - 2 58 Chr. 22 - 32362977 - 32370929 [-] on Build GRCh38
          GenBankmRNA BU569141.1 - 2 134 Chr. 22 - 32360388 - 32371002 [-] on Build GRCh38
          GenBankmRNA BG723497.1 - 2 136 Chr. 22 - 32360422 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY015298.1 - 2 136 Chr. 22 - 32360592 - 32371004 [-] on Build GRCh38
          GenBankmRNA BU584154.1 - 2 58 Chr. 22 - 32363015 - 32370929 [-] on Build GRCh38
          GenBankmRNA CD359353.1 - 2 117 Chr. 22 - 32360312 - 32370990 [-] on Build GRCh38
          GenBankmRNA DB457839.1 - 2 128 Chr. 22 - 32360630 - 32370999 [-] on Build GRCh38
          GenBankmRNA DB447067.1 - 2 128 Chr. 22 - 32360698 - 32370999 [-] on Build GRCh38
          GenBankmRNA BI461189.1 - 2 136 Chr. 22 - 32360588 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY009642.1 - 2 132 Chr. 22 - 32360581 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY028203.1 - 2 132 Chr. 22 - 32360565 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY004848.1 - 2 132 Chr. 22 - 32360571 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY054907.1 - 2 132 Chr. 22 - 32360703 - 32371002 [-] on Build GRCh38
          GenBankmRNA BG771936.1 - 2 136 Chr. 22 - 32360389 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY055002.1 - 2 131 Chr. 22 - 32360704 - 32370999 [-] on Build GRCh38
          GenBankmRNA BG720986.1 - 2 136 Chr. 22 - 32360230 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY034267.1 - 2 132 Chr. 22 - 32360596 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY011074.1 - 2 132 Chr. 22 - 32360626 - 32371002 [-] on Build GRCh38
          GenBankmRNA HY007602.1 - 2 177 Chr. 22 - 32360684 - 32371047 [-] on Build GRCh38
          GenBankmRNA HY002386.1 - 2 132 Chr. 22 - 32360553 - 32371003 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0