Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GGT3P SIRNA Assays ›
          Gene Symbol:
          GGT3P GGT2 LOC102724197 LOC102724823
          Gene Name:
          gamma-glutamyltransferase 3 pseudogene
          gamma-glutamyltransferase 2
          inactive gamma-glutamyltranspeptidase 2-like
          inactive gamma-glutamyltranspeptidase 2
          Gene Aliases:
          GGT3
          GGT, GGT 2
          GGT2
          -
          Chromosome Location:
          Chr.22: 18773689 - 18791961 on Build GRCh38
          Chromosome Location:
          Chr.22: 21207972 - 21282982 on Build GRCh38
          Chromosome Location:
          Chr.22|NT_187386: 65299 - 86956 on Build GRCh38
          Chromosome Location:
          Chr.22|NT_187389: 6 - 18670 on Build GRCh38
          UniGene ID:
          Hs.595809
          Hs.568255
          -
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID n262078

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GGT3P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003267.1 10 1454
          GenBank mRNA BC143607.1 - 6 1200
          Gene Symbol: GGT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005261890.4 XP_005261947.1 6 813
          XM_006724392.3 XP_006724455.1 10 1521
          XM_006724395.3 XP_006724458.1 7 973
          XM_006724396.2 XP_006724459.1 3 351
          XM_011530621.2 XP_011528923.1 3 338
          XM_017029179.1 XP_016884668.1 11 2202
          XM_017029180.1 XP_016884669.1 10 1660
          XM_017029181.1 XP_016884670.1 10 2098
          XM_017029182.1 XP_016884671.1 11 2202
          XM_017029183.1 XP_016884672.1 6 789
          XM_017029184.1 XP_016884673.1 3 361
          XM_017029185.1 XP_016884674.1 12 3064
          XM_017029186.1 XP_016884675.1 15 2665
          GenBank mRNA M30474.1 - 2 147
          Gene Symbol: LOC102724197
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006724921.3 XP_006724984.1 10 1964
          XM_006724924.3 XP_006724987.1 9 1806
          XM_006724928.3 XP_006724991.1 5 1002
          XM_006724929.2 XP_006724992.1 3 351
          XM_011546256.2 XP_011544558.1 11 2098
          XM_017030133.1 XP_016885622.1 11 1529
          XM_017030134.1 XP_016885623.1 11 1532
          XM_017030135.1 XP_016885624.1 11 1957
          XM_017030136.1 XP_016885625.1 12 1999
          XM_017030137.1 XP_016885626.1 12 1996
          XM_017030138.1 XP_016885627.1 12 1519
          XM_017030139.1 XP_016885628.1 12 1522
          XM_017030140.1 XP_016885629.1 13 1989
          XM_017030141.1 XP_016885630.1 13 1986
          XM_017030142.1 XP_016885631.1 11 1830
          XM_017030143.1 XP_016885632.1 10 1965
          XM_017030144.1 XP_016885633.1 6 893
          XM_017030145.1 XP_016885634.1 7 1526
          Gene Symbol: LOC102724823
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017030162.1 XP_016885651.1 11 1968

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydrolase protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          glutathione metabolic process
          glutathione biosynthetic process
          leukotriene biosynthetic process
          xenobiotic metabolic process
          zymogen activation
          gamma-glutamyltransferase activity
          glutathione hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0