Search
Search

CLEC19A| Gene Symbol: CLEC19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256720.1 | NP_001243649.1 | 1 | 126 | ||
| GenBank mRNA | BC118600.1 | - | 1 | 126 | ||
| BC119018.1 | - | 1 | 126 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN273311.1 | - | 1 | 173 | Chr. 16 - 19285739 - 19322145 [+] on Build GRCh38 | |