Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALG2 SIRNA Assays ›
          Gene Symbol:
          ALG2
          Gene Name:
          ALG2, alpha-1,3/1,6-mannosyltransferase
          Gene Aliases:
          CDGIi, CMS14, CMSTA3, NET38, hALPG2
          Chromosome Location:
          Chr.9: 99216425 - 99221964 on Build GRCh38
          UniGene ID:
          Hs.40919
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n263335

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALG2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024532.1 2 501
          GenBank mRNA AK074988.1 - 2 479
          AY358697.1 - 1 86
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AI768004.1 - 2 144 Chr. 9 - 99218594 - 99221607 [-] on Build GRCh38
          GenBankmRNA AW051481.1 - 2 144 Chr. 9 - 99218621 - 99221607 [-] on Build GRCh38
          GenBankmRNA CF132606.1 - 2 268 Chr. 9 - 99218712 - 99221731 [-] on Build GRCh38
          GenBankmRNA DA890710.1 - 2 478 Chr. 9 - 99221011 - 99221940 [-] on Build GRCh38
          GenBankmRNA BP273835.1 - 2 449 Chr. 9 - 99220978 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA430466.1 - 2 450 Chr. 9 - 99218796 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA422820.1 - 2 471 Chr. 9 - 99218808 - 99221923 [-] on Build GRCh38
          GenBankmRNA DA736744.1 - 2 479 Chr. 9 - 99218649 - 99221942 [-] on Build GRCh38
          GenBankmRNA DB059946.1 - 2 449 Chr. 9 - 99221029 - 99221912 [-] on Build GRCh38
          GenBankmRNA BQ641538.1 - 391 Chr. - - [-] on Build
          GenBankmRNA BU594798.1 - 2 482 Chr. 9 - 99218625 - 99221944 [-] on Build GRCh38
          GenBankmRNA HY019575.1 - 2 211 Chr. 9 - 99218635 - 99221548 [-] on Build GRCh38
          GenBankmRNA DB187551.1 - 2 454 Chr. 9 - 99218789 - 99221912 [-] on Build GRCh38
          GenBankmRNA AI278375.1 - 2 144 Chr. 9 - 99218609 - 99221607 [-] on Build GRCh38
          GenBankmRNA DA627469.1 - 2 477 Chr. 9 - 99220989 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA250901.1 - 2 477 Chr. 9 - 99218811 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA714128.1 - 2 459 Chr. 9 - 99218795 - 99221914 [-] on Build GRCh38
          GenBankmRNA DA666759.1 - 2 479 Chr. 9 - 99220988 - 99221942 [-] on Build GRCh38
          GenBankmRNA BP342204.1 - 2 449 Chr. 9 - 99218778 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA928356.1 - 2 457 Chr. 9 - 99218812 - 99221916 [-] on Build GRCh38
          GenBankmRNA DB123779.1 - 2 477 Chr. 9 - 99218812 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA439254.1 - 2 449 Chr. 9 - 99218817 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA904634.1 - 2 478 Chr. 9 - 99220977 - 99221940 [-] on Build GRCh38
          GenBankmRNA CF131378.1 - 2 300 Chr. 9 - 99218633 - 99221763 [-] on Build GRCh38
          GenBankmRNA DB195604.1 - 2 463 Chr. 9 - 99221005 - 99221926 [-] on Build GRCh38
          GenBankmRNA AI863884.1 - 2 144 Chr. 9 - 99218654 - 99221607 [-] on Build GRCh38
          GenBankmRNA DA862318.1 - 2 461 Chr. 9 - 99221029 - 99221916 [-] on Build GRCh38
          GenBankmRNA DR006890.1 - 2 358 Chr. 9 - 99218507 - 99221816 [-] on Build GRCh38
          GenBankmRNA BG773528.1 - 2 296 Chr. 9 - 99218494 - 99221523 [-] on Build GRCh38
          GenBankmRNA AA769683.1 - 2 143 Chr. 9 - 99218552 - 99221606 [-] on Build GRCh38
          GenBankmRNA DA608501.1 - 2 479 Chr. 9 - 99220999 - 99221942 [-] on Build GRCh38
          GenBankmRNA DB000389.1 - 2 478 Chr. 9 - 99220998 - 99221940 [-] on Build GRCh38
          GenBankmRNA AI183323.1 - 2 144 Chr. 9 - 99218659 - 99221607 [-] on Build GRCh38
          GenBankmRNA DA371411.1 - 2 449 Chr. 9 - 99218812 - 99221912 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          dolichol-linked oligosaccharide biosynthetic process
          protein glycosylation in endoplasmic reticulum
          response to calcium ion
          mannosylation
          alpha-1,3-mannosyltransferase activity
          GDP-Man:Man1GlcNAc2-PP-Dol alpha-1,3-mannosyltransferase activity
          protein binding
          protein heterodimerization activity
          protein N-terminus binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hqkpn:80/100.66.73.1:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0