Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALG2 SIRNA Assays ›
          Gene Symbol:
          ALG2
          Gene Name:
          ALG2, alpha-1,3/1,6-mannosyltransferase
          Gene Aliases:
          CDGIi, CMS14, CMSTA3, NET38, hALPG2
          Chromosome Location:
          Chr.9: 99216425 - 99221964 on Build GRCh38
          UniGene ID:
          Hs.40919
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n263336

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALG2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024532.1 2 434
          GenBank mRNA AK074988.1 - 2 412
          AY358697.1 - 1 19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AI768004.1 - 2 77 Chr. 9 - 99218594 - 99221607 [-] on Build GRCh38
          GenBankmRNA AW051481.1 - 2 77 Chr. 9 - 99218621 - 99221607 [-] on Build GRCh38
          GenBankmRNA CF132606.1 - 2 201 Chr. 9 - 99218712 - 99221731 [-] on Build GRCh38
          GenBankmRNA BP273835.1 - 2 382 Chr. 9 - 99220978 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA890710.1 - 2 411 Chr. 9 - 99221011 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA430466.1 - 2 383 Chr. 9 - 99218796 - 99221912 [-] on Build GRCh38
          GenBankmRNA DB059946.1 - 2 382 Chr. 9 - 99221029 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA736744.1 - 2 412 Chr. 9 - 99218649 - 99221942 [-] on Build GRCh38
          GenBankmRNA DA422820.1 - 2 404 Chr. 9 - 99218808 - 99221923 [-] on Build GRCh38
          GenBankmRNA BQ641538.1 - 324 Chr. - - [-] on Build
          GenBankmRNA BU594798.1 - 2 415 Chr. 9 - 99218625 - 99221944 [-] on Build GRCh38
          GenBankmRNA HY019575.1 - 2 144 Chr. 9 - 99218635 - 99221548 [-] on Build GRCh38
          GenBankmRNA DB460341.1 - 2 382 Chr. 9 - 99221049 - 99221912 [-] on Build GRCh38
          GenBankmRNA DB187551.1 - 2 387 Chr. 9 - 99218789 - 99221912 [-] on Build GRCh38
          GenBankmRNA AI278375.1 - 2 77 Chr. 9 - 99218609 - 99221607 [-] on Build GRCh38
          GenBankmRNA DA627469.1 - 2 410 Chr. 9 - 99220989 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA714128.1 - 2 392 Chr. 9 - 99218795 - 99221914 [-] on Build GRCh38
          GenBankmRNA DA250901.1 - 2 410 Chr. 9 - 99218811 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA666759.1 - 2 412 Chr. 9 - 99220988 - 99221942 [-] on Build GRCh38
          GenBankmRNA BP342204.1 - 2 382 Chr. 9 - 99218778 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA928356.1 - 2 390 Chr. 9 - 99218812 - 99221916 [-] on Build GRCh38
          GenBankmRNA DB123779.1 - 2 410 Chr. 9 - 99218812 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA439254.1 - 2 382 Chr. 9 - 99218817 - 99221912 [-] on Build GRCh38
          GenBankmRNA DB195604.1 - 2 396 Chr. 9 - 99221005 - 99221926 [-] on Build GRCh38
          GenBankmRNA CF131378.1 - 2 233 Chr. 9 - 99218633 - 99221763 [-] on Build GRCh38
          GenBankmRNA DA904634.1 - 2 411 Chr. 9 - 99220977 - 99221940 [-] on Build GRCh38
          GenBankmRNA BP246422.1 - 2 393 Chr. 9 - 99221104 - 99221923 [-] on Build GRCh38
          GenBankmRNA DA862318.1 - 2 394 Chr. 9 - 99221029 - 99221916 [-] on Build GRCh38
          GenBankmRNA AI863884.1 - 2 77 Chr. 9 - 99218654 - 99221607 [-] on Build GRCh38
          GenBankmRNA DR006890.1 - 2 291 Chr. 9 - 99218507 - 99221816 [-] on Build GRCh38
          GenBankmRNA BG773528.1 - 2 229 Chr. 9 - 99218494 - 99221523 [-] on Build GRCh38
          GenBankmRNA AA769683.1 - 2 76 Chr. 9 - 99218552 - 99221606 [-] on Build GRCh38
          GenBankmRNA DA608501.1 - 2 412 Chr. 9 - 99220999 - 99221942 [-] on Build GRCh38
          GenBankmRNA DB000389.1 - 2 411 Chr. 9 - 99220998 - 99221940 [-] on Build GRCh38
          GenBankmRNA AI183323.1 - 2 77 Chr. 9 - 99218659 - 99221607 [-] on Build GRCh38
          GenBankmRNA DA371411.1 - 2 382 Chr. 9 - 99218812 - 99221912 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          dolichol-linked oligosaccharide biosynthetic process
          protein glycosylation in endoplasmic reticulum
          response to calcium ion
          mannosylation
          alpha-1,3-mannosyltransferase activity
          GDP-Man:Man1GlcNAc2-PP-Dol alpha-1,3-mannosyltransferase activity
          protein binding
          protein heterodimerization activity
          protein N-terminus binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0