Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL9R SIRNA Assays ›
          Gene Symbol:
          IL9R
          Gene Name:
          interleukin 9 receptor
          Gene Aliases:
          CD129, IL-9R
          Chromosome Location:
          Chr.X: 155997581 - 156013017 on Build GRCh38
          UniGene ID:
          Hs.406228
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n263558

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL9R
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_176786.1 NP_789743.2 2 296
          XM_011531151.2 XP_011529453.1 2 221
          XM_011531152.2 XP_011529454.1 2 221
          XM_011531154.2 XP_011529456.1 2 267
          XM_011531155.2 XP_011529457.1 2 230
          XM_011531156.2 XP_011529458.1 2 228
          XM_011531157.2 XP_011529459.1 2 225
          XM_017029495.1 XP_016884984.1 2 223
          XM_017029496.1 XP_016884985.1 2 231
          XM_017029497.1 XP_016884986.1 2 267
          XM_017029498.1 XP_016884987.1 3 372
          XM_017029499.1 XP_016884988.1 3 294
          XM_017029500.1 XP_016884989.1 2 234
          XM_017029501.1 XP_016884990.1 2 224
          XM_017029503.1 XP_016884992.1 3 297
          XM_017029504.1 XP_016884993.1 2 246
          XM_017029505.1 XP_016884994.1 2 228
          GenBank mRNA AK303178.1 - 2 259
          BC051337.1 - 2 171 6304829
          S71404.1 - 2 221
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BQ057582.1 - 169 Chr. - - [-] on Build
          GenBankmRNA BQ065624.1 - 193 Chr. - - [-] on Build
          GenBankmRNA BQ063826.1 - 2 169 Chr. X - 155997708 - 156005461 [+] on Build GRCh38
          GenBankmRNA BI261351.1 - 2 172 Chr. X - 155997706 - 156005399 [+] on Build GRCh38
          GenBankmRNA BQ058962.1 - 2 168 Chr. X - 155997708 - 156006187 [+] on Build GRCh38
          GenBankmRNA BQ065808.1 - 170 Chr. - - [-] on Build
          GenBankmRNA BQ642675.1 - 2 170 Chr. X - 155997709 - 156005392 [+] on Build GRCh38
          GenBankmRNA BI261391.1 - 2 172 Chr. X - 155997706 - 156005397 [+] on Build GRCh38
          GenBankmRNA BQ057326.1 - 170 Chr. - - [-] on Build
          GenBankmRNA BQ643250.1 - 2 170 Chr. X - 155997711 - 156005465 [+] on Build GRCh38
          GenBankmRNA BQ055765.1 - 2 168 Chr. X - 155997708 - 156005456 [+] on Build GRCh38
          GenBankmRNA BQ059780.1 - 170 Chr. - - [-] on Build
          GenBankmRNA BQ062323.1 - 2 168 Chr. X - 155997708 - 156006176 [+] on Build GRCh38
          GenBankmRNA BQ642408.1 - 2 170 Chr. X - 155997711 - 156005456 [+] on Build GRCh38
          GenBankmRNA BQ058298.1 - 2 168 Chr. X - 155997708 - 156006123 [+] on Build GRCh38
          GenBankmRNA BQ064101.1 - 2 170 Chr. X - 155997711 - 156005422 [+] on Build GRCh38
          GenBankmRNA BQ065789.1 - 2 170 Chr. X - 155997711 - 156005404 [+] on Build GRCh38
          GenBankmRNA BQ064927.1 - 2 167 Chr. X - 155997711 - 156005470 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          cell proliferation
          interleukin-9-mediated signaling pathway
          positive regulation of protein targeting to mitochondrion
          interleukin-9 receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0