Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL9R SIRNA Assays ›
          Gene Symbol:
          IL9R
          Gene Name:
          interleukin 9 receptor
          Gene Aliases:
          CD129, IL-9R
          Chromosome Location:
          Chr.X: 155997581 - 156013017 on Build GRCh38
          UniGene ID:
          Hs.406228
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n263559

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL9R
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_176786.1 NP_789743.2 2 246
          XM_011531151.2 XP_011529453.1 2 171
          XM_011531152.2 XP_011529454.1 2 171
          XM_011531154.2 XP_011529456.1 2 217
          XM_011531155.2 XP_011529457.1 2 180
          XM_011531156.2 XP_011529458.1 2 178
          XM_011531157.2 XP_011529459.1 2 175
          XM_017029495.1 XP_016884984.1 2 173
          XM_017029496.1 XP_016884985.1 2 181
          XM_017029497.1 XP_016884986.1 2 217
          XM_017029498.1 XP_016884987.1 3 322
          XM_017029499.1 XP_016884988.1 3 244
          XM_017029500.1 XP_016884989.1 2 184
          XM_017029501.1 XP_016884990.1 2 174
          XM_017029503.1 XP_016884992.1 3 247
          XM_017029504.1 XP_016884993.1 2 196
          XM_017029505.1 XP_016884994.1 2 178
          GenBank mRNA AK303178.1 - 2 209
          BC051337.1 - 2 121 6304829
          S71404.1 - 2 171
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BQ057582.1 - 119 Chr. - - [-] on Build
          GenBankmRNA BQ065624.1 - 143 Chr. - - [-] on Build
          GenBankmRNA BQ063826.1 - 2 119 Chr. X - 155997708 - 156005461 [+] on Build GRCh38
          GenBankmRNA BI261351.1 - 2 122 Chr. X - 155997706 - 156005399 [+] on Build GRCh38
          GenBankmRNA BQ058962.1 - 2 118 Chr. X - 155997708 - 156006187 [+] on Build GRCh38
          GenBankmRNA BQ065808.1 - 120 Chr. - - [-] on Build
          GenBankmRNA BQ642675.1 - 2 120 Chr. X - 155997709 - 156005392 [+] on Build GRCh38
          GenBankmRNA BI261391.1 - 2 122 Chr. X - 155997706 - 156005397 [+] on Build GRCh38
          GenBankmRNA BQ057326.1 - 120 Chr. - - [-] on Build
          GenBankmRNA BQ643250.1 - 2 120 Chr. X - 155997711 - 156005465 [+] on Build GRCh38
          GenBankmRNA BQ055765.1 - 2 118 Chr. X - 155997708 - 156005456 [+] on Build GRCh38
          GenBankmRNA BQ062323.1 - 2 118 Chr. X - 155997708 - 156006176 [+] on Build GRCh38
          GenBankmRNA BQ059780.1 - 120 Chr. - - [-] on Build
          GenBankmRNA BQ642408.1 - 2 120 Chr. X - 155997711 - 156005456 [+] on Build GRCh38
          GenBankmRNA BQ058298.1 - 2 118 Chr. X - 155997708 - 156006123 [+] on Build GRCh38
          GenBankmRNA BQ064101.1 - 2 120 Chr. X - 155997711 - 156005422 [+] on Build GRCh38
          GenBankmRNA BQ065789.1 - 2 120 Chr. X - 155997711 - 156005404 [+] on Build GRCh38
          GenBankmRNA BQ064927.1 - 2 117 Chr. X - 155997711 - 156005470 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          cell proliferation
          interleukin-9-mediated signaling pathway
          positive regulation of protein targeting to mitochondrion
          interleukin-9 receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0