Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FBXO22-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003136.1 | 1 | 110 | |||
GenBank mRNA | AA843761.1 | - | 1 | 114 | ||
AA931120.1 | - | 1 | 115 | |||
AI868357.1 | - | 1 | 114 |