Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242336.1 | NP_001229265.1 | 1 | 315 | ||
GenBank mRNA | AL037715.1 | - | 1 | 284 | ||
BC026244.1 | - | 1 | 275 | 4697443 |