Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MGC72080 SIRNA Assays ›
          Gene Symbol:
          MGC72080
          Gene Name:
          MGC72080 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 97966596 - 97972326 on Build GRCh38
          UniGene ID:
          Hs.655977
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n264721

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MGC72080
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002822.3 1, 2 193
          GenBank mRNA BC029615.1 - 1, 2 203
          BC042035.1 - 1, 2 180
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA HY148228.1 - 1, 2 193 Chr. 7 - 97966842 - 97972326 [-] on Build GRCh38
          GenBankmRNA CB995592.1 - 1, 2 237 Chr. 7 - 97966608 - 97972344 [-] on Build GRCh38
          GenBankmRNA BG032285.1 - 1, 2 172 Chr. 7 - 97966601 - 97972305 [-] on Build GRCh38
          GenBankmRNA BQ435340.1 - 220 Chr. - - [-] on Build
          GenBankmRNA BG831490.1 - 1, 2 210 Chr. 7 - 97966590 - 97972343 [-] on Build GRCh38
          GenBankmRNA AW361749.1 - 1, 2 80 Chr. 7 - 97966832 - 97972213 [-] on Build GRCh38
          GenBankmRNA HY022048.1 - 1, 2 200 Chr. 7 - 97966853 - 97972332 [-] on Build GRCh38
          GenBankmRNA BG751054.1 - 1, 2 196 Chr. 7 - 97966370 - 97972328 [-] on Build GRCh38
          GenBankmRNA BE893680.1 - 1, 2 140 Chr. 7 - 97966600 - 97972273 [-] on Build GRCh38
          GenBankmRNA HY157451.1 - 1, 2 193 Chr. 7 - 97966848 - 97972326 [-] on Build GRCh38
          GenBankmRNA BQ883317.1 - 165 Chr. - - [-] on Build
          GenBankmRNA BU158741.1 - 1, 2 175 Chr. 7 - 97966611 - 97972307 [-] on Build GRCh38
          GenBankmRNA CD675601.1 - 1, 2 179 Chr. 7 - 97966672 - 97972277 [-] on Build GRCh38
          GenBankmRNA HY084818.1 - 1, 2 176 Chr. 7 - 97966812 - 97972309 [-] on Build GRCh38
          GenBankmRNA DN831471.1 - 1, 2 35 Chr. 7 - 97966691 - 97972168 [-] on Build GRCh38
          GenBankmRNA HY143129.1 - 1, 2 220 Chr. 7 - 97966909 - 97972352 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xhq5n:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0