Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023918.2 | 2 | 254 | |||
NR_023919.2 | 2 | 212 | ||||
NR_109759.1 | 2 | 208 | ||||
NR_109760.1 | 2 | 209 | ||||
NR_109761.1 | 3 | 329 | ||||
NR_109762.1 | 3 | 333 | ||||
GenBank mRNA | AF161506.1 | - | 2 | 182 | ||
BC016053.1 | - | 2 | 167 | |||
BG432520.1 | - | 3 | 323 | |||
BX509008.1 | - | 2 | 209 | |||
CA489192.1 | - | 2 | 231 | |||
H87321.1 | - | 3 | 328 |