Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PGM5P2 SIRNA Assays ›
          Gene Symbol:
          PGM5P2 HELLPAR
          Gene Name:
          phosphoglucomutase 5 pseudogene 2
          HELLP associated long non-coding RNA
          Gene Aliases:
          -
          LINC-HELLP, lncHELLP
          Chromosome Location:
          Chr.9: 41007011 - 41074625 on Build GRCh38
          Chromosome Location:
          -
          UniGene ID:
          Hs.571593
          Hs.160562
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n265981

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PGM5P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002836.2 8, 9 1463
          GenBank mRNA BC025351.1 - 8, 9 1455
          Gene Symbol: HELLPAR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA JX088243.1 - 1 115144
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA N70045.1 - 3 256 Chr. 12 - 51328571 - 51345872 [-] on Build GRCh38
          GenBankmRNA AF279660.3 - 6 188222 Chr. 13 - 49996975 - 50344526 [+] on Build GRCh38
          GenBankmRNA BX954043.1 - 1 273 Chr. 6 - 19496061 - 19766208 [-] on Build GRCh38
          GenBankmRNA AL580086.3 - 4 644 Chr. 9 - 40993279 - 41014097 [-] on Build GRCh38
          GenBankmRNA DA100091.1 - 1 258 Chr. 15 - 43650728 - 43651271 [-] on Build GRCh38
          GenBankmRNA BX953697.1 - 1 419 Chr. 3 - 159396872 - 159397356 [+] on Build GRCh38
          GenBankmRNA AA774117.1 - 1 58 Chr. 6 - 133146985 - 133147395 [+] on Build GRCh38
          GenBankmRNA AA807733.1 - 1 54 Chr. 3 - 88508011 - 88508227 [+] on Build GRCh38
          GenBankmRNA BG012085.1 - 1 324 Chr. 12 - 39699952 - 39979684 [-] on Build GRCh38
          GenBankmRNA DB288875.1 - 1 255 Chr. 16 - 47183619 - 47184084 [-] on Build GRCh38
          GenBankmRNA DB356793.1 - 1 257 Chr. 19 - 12009503 - 12010068 [+] on Build GRCh38
          GenBankmRNA BU676599.1 - 66 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0