Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PMCHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003921.1 | 4 | 481 | |||
GenBank mRNA | AY008407.1 | - | 5 | 559 | ||
AY008408.1 | - | 5 | 563 | |||
AY008409.1 | - | 4 | 481 | |||
AY008410.1 | - | 4 | 519 | |||
AY008411.1 | - | 4 | 1740 | |||
EU921424.1 | - | 5 | 395 | |||
EU921426.1 | - | 5 | 466 | |||
EU921427.1 | - | 5 | 506 | |||
EU921428.1 | - | 4 | 389 | |||
EU921429.1 | - | 5 | 466 | |||
EU921430.1 | - | 6 | 662 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX091674.1 | - | 4 | 407 | Chr. 5 - 22142464 - 22152246 [+] on Build GRCh38 |