Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JPX SIRNA Assays ›
          Gene Symbol:
          JPX
          Gene Name:
          JPX transcript, XIST activator (non-protein coding)
          Gene Aliases:
          DCBALD06, ENOX, LINC00183, NCRNA00183
          Chromosome Location:
          Chr.X: 73944324 - 74070384 on Build GRCh38
          UniGene ID:
          Hs.648316
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n266314

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JPX
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024582.1
          1 100
          GenBank mRNA AK056172.1 -
          1 82
          AV714079.1 - LIT1011
          Jpx
          85
          BC071776.1 -
          1 96
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BG504771.1 - 1 79 Chr. X - 73944346 - 73947528 [+] on Build GRCh38
          GenBankmRNA DA979750.1 - 1 73 Chr. X - 73944351 - 73947352 [+] on Build GRCh38
          GenBankmRNA BM548256.1 - 132 Chr. - - [-] on Build
          GenBankmRNA BP872745.1 - 1 79 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP871836.1 - 1 78 Chr. X - 73944346 - 73947297 [+] on Build GRCh38
          GenBankmRNA CN270680.1 - 1 65 Chr. X - 73944359 - 73999372 [+] on Build GRCh38
          GenBankmRNA AW963219.1 - 72 Chr. - - [-] on Build
          GenBankmRNA AV705096.1 - 1 35 Chr. X - 73944389 - 74004413 [+] on Build GRCh38
          GenBankmRNA BU934969.1 - 1 80 Chr. X - 73944346 - 73999254 [+] on Build GRCh38
          GenBankmRNA Z44786.1 - 67 Chr. X - 123628779 - 123628794 [-] on Build GRCh38
          GenBankmRNA AL711317.1 - 1 99 Chr. X - 73944324 - 73999368 [+] on Build GRCh38
          GenBankmRNA BG612854.1 - 1 81 Chr. X - 73944343 - 73999478 [+] on Build GRCh38
          GenBankmRNA HY065667.1 - 1 86 Chr. X - 73944340 - 74010499 [+] on Build GRCh38
          GenBankmRNA CB993733.1 - 109 Chr. - - [-] on Build
          GenBankmRNA BP872499.1 - 1 100 Chr. X - 73944324 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP873595.1 - 1 82 Chr. X - 73944342 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP872256.1 - 1 79 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA DB255033.1 - 1 86 Chr. X - 73944339 - 73947366 [+] on Build GRCh38
          GenBankmRNA F12346.1 - 41 Chr. - - [-] on Build
          GenBankmRNA CF129475.1 - 1 65 Chr. X - 73944359 - 74004553 [+] on Build GRCh38
          GenBankmRNA DA363254.1 - 1 99 Chr. X - 73944326 - 73997077 [+] on Build GRCh38
          GenBankmRNA DA106100.1 - 1 82 Chr. X - 73944342 - 74004417 [+] on Build GRCh38
          GenBankmRNA CN263834.1 - 1 68 Chr. X - 73944356 - 73999314 [+] on Build GRCh38
          GenBankmRNA HY103315.1 - 1 82 Chr. X - 73944342 - 73947268 [+] on Build GRCh38
          GenBankmRNA BM810643.1 - 55 Chr. - - [-] on Build
          GenBankmRNA BU164379.1 - 1 75 Chr. X - 73944349 - 73998832 [+] on Build GRCh38
          GenBankmRNA BP871739.1 - 1 79 Chr. X - 73944345 - 74004345 [+] on Build GRCh38
          GenBankmRNA BP872588.1 - 1 79 Chr. X - 73944345 - 74004345 [+] on Build GRCh38
          GenBankmRNA BP871919.1 - 1 80 Chr. X - 73944344 - 73999613 [+] on Build GRCh38
          GenBankmRNA DA841100.1 - 1 79 Chr. X - 73944345 - 73947361 [+] on Build GRCh38
          GenBankmRNA BG613915.1 - 1 83 Chr. X - 73944342 - 73999860 [+] on Build GRCh38
          GenBankmRNA AL708895.1 - 1 87 Chr. X - 73944338 - 73947333 [+] on Build GRCh38
          GenBankmRNA BU537716.1 - 1 94 Chr. X - 73944333 - 73947334 [+] on Build GRCh38
          GenBankmRNA BP871940.1 - 1 79 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA DA331725.1 - 1 82 Chr. X - 73944342 - 74004540 [+] on Build GRCh38
          GenBankmRNA BP873466.1 - 1 100 Chr. X - 73944324 - 73999111 [+] on Build GRCh38
          GenBankmRNA BP871783.1 - 1 79 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA R19849.1 - 1 63 Chr. X - 73944385 - 74047792 [+] on Build GRCh38
          GenBankmRNA HY088843.1 - 1 86 Chr. X - 73944340 - 74010567 [+] on Build GRCh38
          GenBankmRNA HY172225.1 - 1 83 Chr. X - 73944342 - 73999019 [+] on Build GRCh38
          GenBankmRNA BP872277.1 - 1 100 Chr. X - 73944324 - 73999592 [+] on Build GRCh38
          GenBankmRNA BG435566.1 - 1 97 Chr. X - 73944328 - 74069325 [+] on Build GRCh38
          GenBankmRNA DA103086.1 - 1 81 Chr. X - 73944343 - 74004499 [+] on Build GRCh38
          GenBankmRNA BU589636.1 - 1 81 Chr. X - 73944346 - 73944948 [+] on Build GRCh38
          GenBankmRNA DA427047.1 - 1 98 Chr. X - 73944326 - 73947342 [+] on Build GRCh38
          GenBankmRNA CB991754.1 - 109 Chr. - - [-] on Build
          GenBankmRNA DB240432.1 - 1 78 Chr. X - 73944346 - 73947385 [+] on Build GRCh38
          GenBankmRNA DA881370.1 - 1 77 Chr. X - 73944347 - 73999014 [+] on Build GRCh38
          GenBankmRNA DA679611.1 - 1 82 Chr. X - 73944342 - 73999105 [+] on Build GRCh38
          GenBankmRNA C16093.1 - 1 18 Chr. 3 - 71682388 - 71682656 [-] on Build GRCh38
          GenBankmRNA DA731241.1 - 1 98 Chr. X - 73944326 - 73998845 [+] on Build GRCh38
          GenBankmRNA BP873513.1 - 1 79 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA T32551.1 - 71 Chr. 15 - 42410703 - 42410722 [-] on Build GRCh38
          GenBankmRNA BP872419.1 - 1 100 Chr. X - 73944324 - 74004351 [+] on Build GRCh38
          GenBankmRNA BF686802.1 - 63 Chr. - - [-] on Build
          GenBankmRNA DA700446.1 - 1 82 Chr. X - 73944342 - 73947318 [+] on Build GRCh38
          GenBankmRNA DB250056.1 - 1 78 Chr. X - 73944346 - 73999096 [+] on Build GRCh38
          GenBankmRNA BG427198.1 - 1 83 Chr. X - 73944342 - 73995436 [+] on Build GRCh38
          GenBankmRNA DW450219.1 - 1 95 Chr. X - 73944342 - 73944645 [+] on Build GRCh38
          GenBankmRNA DW466961.1 - 1 97 Chr. X - 73944340 - 73994872 [+] on Build GRCh38
          GenBankmRNA BP871862.1 - 1 84 Chr. X - 73944340 - 73947374 [+] on Build GRCh38
          GenBankmRNA BP873504.1 - 1 76 Chr. X - 73944356 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP872587.1 - 1 80 Chr. X - 73944344 - 74004351 [+] on Build GRCh38
          GenBankmRNA HY081032.1 - 1 85 Chr. X - 73944339 - 73947244 [+] on Build GRCh38
          GenBankmRNA BP873185.1 - 1 80 Chr. X - 73944344 - 74004355 [+] on Build GRCh38
          GenBankmRNA H84526.1 - 60 Chr. - - [-] on Build
          GenBankmRNA BU599790.1 - 1 85 Chr. X - 73944340 - 74004542 [+] on Build GRCh38
          GenBankmRNA BP872964.1 - 1 79 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA BF797301.1 - 1 67 Chr. X - 73944357 - 73999368 [+] on Build GRCh38
          GenBankmRNA R27273.1 - 64 Chr. - - [-] on Build
          GenBankmRNA BP872836.1 - 1 80 Chr. X - 73944344 - 74004351 [+] on Build GRCh38
          GenBankmRNA DB210310.1 - 1 92 Chr. X - 73944332 - 73999117 [+] on Build GRCh38
          GenBankmRNA DA711535.1 - 1 83 Chr. X - 73944342 - 73947373 [+] on Build GRCh38
          GenBankmRNA CB998281.1 - 109 Chr. - - [-] on Build
          GenBankmRNA BP872490.1 - 1 80 Chr. X - 73944344 - 74004345 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq5tn:80/100.66.79.35:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0