Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LINC00472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026807.2 | 4 | 2147 | |||
NR_121612.1 | 4 | 2198 | ||||
NR_121613.1 | 3 | 2132 | ||||
NR_121614.1 | 3 | 2081 | ||||
GenBank mRNA | AK023251.1 | - | 4 | 2147 | ||
BM720568.1 | - | 1 | 547 |