Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD20A11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027270.1 | 1 | 547 | |||
GenBank mRNA | AF426257.1 | - | 1 | 547 | ||
BC043199.1 | - | 1 | 547 |
Gene Symbol: ANKRD20A5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040113.1 | 1 | 305 | |||
GenBank mRNA | BC022023.1 | - | 1 | 305 | 4824658 | |
BC127726.1 | - | 1 | 84 | 40127384 | ||
BP201899.1 | - | 1 | 33 | |||
DB446908.1 | - | 1 | 280 |