Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYO15B SIRNA Assays ›
          Gene Symbol:
          MYO15B
          Gene Name:
          myosin XVB
          Gene Aliases:
          MYO15BP
          Chromosome Location:
          Chr.17: 75587545 - 75626850 on Build GRCh38
          UniGene ID:
          Hs.390817
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n268131

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYO15B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001309242.1 NP_001296171.1 41 6799
          XM_017025120.1 XP_016880609.1 43 6506
          XM_017025121.1 XP_016880610.1 43 6491
          XM_017025122.1 XP_016880611.1 43 6470
          XM_017025123.1 XP_016880612.1 43 6508
          XM_017025124.1 XP_016880613.1 42 6380
          XM_017025125.1 XP_016880614.1 41 6229
          XM_017025126.1 XP_016880615.1 38 5971
          XM_017025127.1 XP_016880616.1 41 5814
          XM_017025128.1 XP_016880617.1 42 5560
          XM_017025129.1 XP_016880618.1 41 4792
          XM_017025130.1 XP_016880619.1 40 4748
          XM_017025131.1 XP_016880620.1 39 5187
          XM_017025132.1 XP_016880621.1 38 5128
          XM_017025133.1 XP_016880622.1 36 4653
          XM_017025134.1 XP_016880623.1 37 5014
          XM_017025135.1 XP_016880624.1 31 3670
          XM_017025136.1 XP_016880625.1 31 4019
          XM_017025137.1 XP_016880626.1 28 3564
          XM_017025138.1 XP_016880627.1 23 3060
          XM_017025139.1 XP_016880628.1 24 2861
          XM_017025140.1 XP_016880629.1 21 2674
          XM_017025143.1 XP_016880632.1 16 1951
          XM_017025144.1 XP_016880633.1 37 6434
          GenBank mRNA AB058686.1 - 31 4906
          AF418286.1 - 37 6816
          AF418290.1 - 36 7227
          AF418292.1 - 17 2015
          AF418293.1 - 17 2015
          AK025259.1 - 967
          AK097896.1 - 9 1275
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB138051.1 - 2 134 Chr. 17 - 75617270 - 75619805 [+] on Build GRCh38
          GenBankmRNA BF364690.1 - 2 270 Chr. 17 - 75617141 - 75618141 [+] on Build GRCh38
          GenBankmRNA CR984271.1 - 1 85 Chr. 17 - 75617824 - 75620358 [+] on Build GRCh38
          GenBankmRNA EL954657.1 - 3 389 Chr. 17 - 75616913 - 75619386 [+] on Build GRCh38
          GenBankmRNA DA316903.1 - 2 279 Chr. 17 - 75617125 - 75619454 [+] on Build GRCh38
          GenBankmRNA BI766913.1 - 451 Chr. - - [-] on Build
          GenBankmRNA CR986155.1 - 2 237 Chr. 17 - 75617167 - 75619796 [+] on Build GRCh38
          GenBankmRNA DA304564.1 - 1 95 Chr. 17 - 75617814 - 75619957 [+] on Build GRCh38
          GenBankmRNA DB134005.1 - 1 66 Chr. 17 - 75617843 - 75619964 [+] on Build GRCh38
          GenBankmRNA DA109947.1 - 1 430 Chr. 17 - 75617496 - 75619182 [+] on Build GRCh38
          GenBankmRNA DA428235.1 - 2 209 Chr. 17 - 75617580 - 75619565 [+] on Build GRCh38
          GenBankmRNA BI772088.1 - 351 Chr. - - [-] on Build
          GenBankmRNA DA126806.1 - 4 481 Chr. 17 - 75616712 - 75619177 [+] on Build GRCh38
          GenBankmRNA DB138105.1 - 2 134 Chr. 17 - 75617270 - 75619805 [+] on Build GRCh38
          GenBankmRNA BM804816.1 - 5 697 Chr. 17 - 75616419 - 75619471 [+] on Build GRCh38
          GenBankmRNA BY798869.2 - 1 103 Chr. 17 - 75617810 - 75619955 [+] on Build GRCh38
          GenBankmRNA BI769659.1 - 4 491 Chr. 17 - 75616699 - 75619377 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2gfbh:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0