Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYO15B SIRNA Assays ›
          Gene Symbol:
          MYO15B
          Gene Name:
          myosin XVB
          Gene Aliases:
          MYO15BP
          Chromosome Location:
          Chr.17: 75587545 - 75626850 on Build GRCh38
          UniGene ID:
          Hs.390817
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n268133

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYO15B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001309242.1 NP_001296171.1 36 5973
          XM_017025120.1 XP_016880609.1 38 5680
          XM_017025121.1 XP_016880610.1 38 5665
          XM_017025122.1 XP_016880611.1 38 5644
          XM_017025123.1 XP_016880612.1 38 5682
          XM_017025124.1 XP_016880613.1 37 5554
          XM_017025125.1 XP_016880614.1 36 5403
          XM_017025126.1 XP_016880615.1 33 5145
          XM_017025127.1 XP_016880616.1 36 4988
          XM_017025128.1 XP_016880617.1 37 4734
          XM_017025129.1 XP_016880618.1 36 3966
          XM_017025130.1 XP_016880619.1 35 3922
          XM_017025131.1 XP_016880620.1 34 4361
          XM_017025132.1 XP_016880621.1 33 4302
          XM_017025133.1 XP_016880622.1 31 3827
          XM_017025134.1 XP_016880623.1 32 4188
          XM_017025135.1 XP_016880624.1 26 2844
          XM_017025136.1 XP_016880625.1 26 3193
          XM_017025137.1 XP_016880626.1 23 2738
          XM_017025138.1 XP_016880627.1 18 2234
          XM_017025139.1 XP_016880628.1 19 2035
          XM_017025140.1 XP_016880629.1 16 1848
          XM_017025143.1 XP_016880632.1 11 1125
          XM_017025144.1 XP_016880633.1 32 5608
          GenBank mRNA AB058686.1 - 26 4080
          AF418289.1 - 32 5608
          AF418290.1 - 31 6401
          AF418292.1 - 12 1190
          AF418293.1 - 12 1190
          AK097896.1 - 4 449
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BQ374677.1 - 2 177 Chr. 17 - 75615767 - 75616685 [+] on Build GRCh38
          GenBankmRNA DA933596.1 - 6 545 Chr. 17 - 75614833 - 75616139 [+] on Build GRCh38
          GenBankmRNA DB149783.1 - 4 439 Chr. 17 - 75615247 - 75616350 [+] on Build GRCh38
          GenBankmRNA DA257555.1 - 4 401 Chr. 17 - 75615265 - 75616381 [+] on Build GRCh38
          GenBankmRNA BQ374676.1 - 2 167 Chr. 17 - 75615777 - 75616684 [+] on Build GRCh38
          GenBankmRNA DA107653.1 - 1 23 Chr. 17 - 75616103 - 75616734 [+] on Build GRCh38
          GenBankmRNA BX504202.1 - 4 427 Chr. 17 - 75615259 - 75616410 [+] on Build GRCh38
          GenBankmRNA DB106255.1 - 4 449 Chr. 17 - 75615239 - 75616392 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0