Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NXF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028089.1 | 19 | 1842 | |||
GenBank mRNA | AJ277654.1 | - | 19 | 1842 | ||
AJ277655.1 | - | 19 | 1817 | |||
AJ277656.1 | - | 19 | 1753 | |||
AJ277657.1 | - | 17 | 1628 | |||
AJ277658.1 | - | 17 | 1747 |